Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
form
liquid
manufacturer/tradename
USP
application(s)
USP Biologics
pharmaceutical (small molecule)
format
single component solution
storage temp.
2-8°C
General description
Bovine Serum Albumin (BSA) is a globular protein derived from bovine blood plasma. It is widely utilized as a standard in protein quantitation assays due to its stability and lack of interference with the assay components.
The USP BSA for Protein Quantitation standard serves as a calibration reference in assays such as the Bradford and Bicinchoninic Acid (BCA) assays, ensuring accurate and reproducible measurement of protein concentrations in various research and clinical applications.
The USP biologics proteins category provides critical standards and reference materials necessary for the development and manufacturing of therapeutic proteins. These proteins, which can be naturally derived or genetically engineered, are essential in treating various diseases and conditions, ensuring that they meet quality, safety, and efficacy standards.
The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.
Application
Also used to prepare standard and suitability standard solutions during the determination of total protein in pharmaceutical drug substances and products according to the United States Pharmacopeia (USP) general chapter.
Biochem/physiol Actions
Preparation Note
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade - | grade - | grade - |
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename Probumin® | manufacturer/tradename Probumin® |
| format single component solution | format - | format - | format - |
| form liquid | form lyophilized powder | form lyophilized powder | form lyophilized powder, solution (30%) |
| application(s) USP Biologics | application(s) hematology | application(s) - | application(s) - |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1076192-5X1ML | 04061837842580 |


