G005800
GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on SUPELCOWAX 10
suitable for GC
About This Item
Recommended Products
technique(s)
gas chromatography (GC): suitable
test parameters
column: SUPELCOWAX 10, 30 m x 0.32 mm I.D., 0.25 μm (24082)
oven: 50°C (1 min), 5 °C/min to 225 °C (5 min)
inj. temp.: 250 °C
detector: FID, 250 °C
carrier gas: helium, 25 cm/sec (set at 200 °C)
injection: 1 μL 100:1 split
liner: 4 mm I.D., cup style
sample: bacterial methyl ester standards in methyl caproate, 10 mg/mL (47080-U)
application(s)
clinical
Legal Information
analytical column
standard
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service