59562
SUPELCOSIL™ LC-8 (5 µm) HPLC Columns
L × I.D. 2 cm × 4 mm Supelguard Guard Cartridge pkg of 2 ea, Guard Cartridge holder required for use
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115700
eCl@ss:
32110501
Recommended Products
Product Name
SUPELCOSIL™ LC-8 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Agency
suitable for USP L7
Quality Level
packaging
pkg of 2 ea
technique(s)
HPLC: suitable
L × I.D.
2 cm × 4 mm
matrix active group
C8 (octyl) phase
particle size
5 μm
pore size
120 Å
application(s)
food and beverages
separation technique
reversed phase
Looking for similar products? Visit Product Comparison Guide
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thomas Borch et al.
Journal of chromatography. A, 1022(1-2), 83-94 (2004-02-03)
Explosives such as 2,4,6-trinitrotoluene (TNT), octahydro-1,3,5,7-tetranitro-1,3,5,7-tetrazocine (HMX), and hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX) are widely distributed environmental contaminants. Complete chromatographic separation is necessary in order to accurately determine and quantify explosives and their degradation products in environmental samples and in (bio)transformation studies. The
Jadwiga Piwowarska et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 805(1), 1-5 (2004-04-29)
A high-performance liquid chromatographic method is described for determination of lidocaine (2-(dietyloamino)-N-(2,6-dimetylofenylo) acetamid) and its metabolite, monoethylglycine xylidide (MEGX), in human serum containing various concentration of bilirubin. Lidocaine and its metabolite were extracted from human serum using dichloromethane. After separation
P K Kunicki
Journal of chromatography. B, Biomedical sciences and applications, 750(1), 41-49 (2001-02-24)
A HPLC-UV determination of clobazam and N-desmethylclobazam in human serum and urine is presented. After simple liquid-liquid extraction with dichloromethane the compounds and an internal standard diazepam were separated on a Supelcosil LC-8-DB column at ambient temperature under isocratic conditions
F Belal et al.
Journal of pharmaceutical and biomedical analysis, 24(3), 335-342 (2001-02-24)
A simple, rapid and sensitive HPLC method has been developed for the simultaneous determination of ramipril and hydrochlorothiazide in their dosage forms. Acetonitrile: sodium perchlorate solution (0.1 M) adjusted to pH 2.5+/-0.2 with phosphoric acid (46:54 v/v), was used as
J H Nichols et al.
Clinical chemistry, 40(7 Pt 1), 1312-1316 (1994-07-01)
This automated assay determines the concentration of the antidepressant fluoxetine (Prozac) and its active metabolite norfluoxetine in serum by reversed-phase HPLC with spectrophotometric detection. Extraction, injection, and quantification are performed by the Gilson Aspec automated sample handler. The patient's specimen
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service