Skip to Content
Merck
All Photos(2)

Documents

57343-U

Supelco

SPME Fiber Assembly

greener alternative

CAR/PDMS, for use with autosampler, Nitinol-core (NIT), 23 ga, 75 μm

Synonym(s):

Carboxen/PDMS SPME fiber assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

product name

SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler

material

fused silica fiber
plain black hub

needle size

23 ga

packaging

pkg of 3 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

75 μm (CAR/PDMS)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

CAR/PDMS coating

application(s)

food and beverages

compatibility

(For gasses and low molecular weight compounds (MW 30 - 225).)
for use with autosampler

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

For Gases and Low Molecular Weight Compounds (MW 30-225)
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Footnote

This SPME fiber assembly is also available with a Nitinol (NIT) core as Cat. No. 57907-U, providing enhanced fiber reproducibility, greater fiber durability, maintained selectivity for method consistency, and core inertness to ensure analyte stability. See more at SigmaAldrich.com/NITSPME .

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Talon, R., Montel, M.
Applied SPME, 364-371 (1999)
Pawliszyn, Janusz
Solid Phase Microextraction: Theory and Practice, 264-264 (1997)
Pawliszyn, Janusz
Applications of Solid Phase Microextraction, 674-674 (1999)
Pawliszyn, Janusz
Handbook of Solid Phase Microextraction, 410-410 (2009)
Edited by Sue Ann Scheppers Wercinski
Solid Phase Microextraction: A Practical Guide, 257-257 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service