23446-U
ORBO™-93 Sorbent Tube
ORBO tube I.D. × L 6 mm × 95 mm, Bed A 180 mg, Bed B 90 mg, 60/80 mesh, pkg of 25 ea
Synonym(s):
Carboxen® 1000
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product line
ORBO™
Quality Level
composition
Bed A, 180 mg
Bed B, 90 mg
packaging
pkg of 25 ea
technique(s)
active air sampling: suitable
ORBO tube I.D. × L
6 mm × 95 mm
particle size
60/80 mesh
application(s)
air monitoring
environmental
industrial hygiene
General description
ORBO sorbent tubes (matrix Carboxen® 1000, ORBO tube I.D. × L 6 mm × 95 mm, Bed B 90 mg, Bed A 180 mg, 60/80 mesh) contain two beds of the same selective adsorbent separated by glass wool or foam, for gas and vapor sampling. The dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
Legal Information
Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany
ORBO is a trademark of Sigma-Aldrich Co. LLC
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of agricultural and food chemistry, 52(17), 5431-5435 (2004-08-19)
A rapid and automated solid phase microextraction (SPME) stable isotope dilution gas chromatography/mass spectrometry (GC-MS) method for 2-methoxy-3-isobutylpyrazine (MIBP) quantification in red wine was developed. Wines with 30% (w/v) NaCl and 2-methoxy-(2)H(3)-3-isobutylpyrazine internal standard were sampled with a 2 cm
Journal of chromatography. A, 1238, 15-21 (2012-04-13)
Trihalomethanes are predominantly formed during disinfection of water via reactions of the oxidant with natural organic matter. Even though chlorinated and brominated trihalomethanes are the most widespread organic contaminants in drinking water, when iodide is present in raw water iodinated
Journal of chromatography. A, 1017(1-2), 141-149 (2003-10-31)
A new method was developed for the determination of 2-furfural (2-F) and 5-methylfurfural (5-MF), two products of Maillard reaction in vinegar, with head-space solid-phase microextraction (HS-SPME) coupled to gas chromatography-mass spectrometry (GC-MS). A divinylbenzene (DVB)/carboxen (CAR)/polydimethylsiloxane (PDMS) fibre was used
Journal of chromatography. A, 991(1), 1-11 (2003-04-22)
The solid-phase microextraction (SPME) device was used as a time-weighted average (TWA) sampler for ethylene oxide. Carboxen/polydimethylsiloxane (CAR/PDMS) fiber was used and hydrogen bromide (HBr) was loaded onto the fiber. The SPME fiber assembly was then inserted into PTFE tubing
Chemistry & biodiversity, 8(1), 95-114 (2011-01-25)
Headspace solid-phase microextraction (HS-SPME) coupled with GC/FID and GC/MS was applied for the first time in the analysis of the volatile fraction of an Ephedra species. Notably, six Italian populations (Marche, Abruzzo, and Sardinia) of Ephedra nebrodensis subsp. nebrodensis, covering
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service