Key Documents
Z600768
Hellma® fluorescence cuvettes, semi Micro
High Performance Quartz Glass, spectral range 200-2,500 nm, pathlength 10x4 mm, chamber volume 1400 μL
Synonym(s):
Semi-Micro Fluorescence Cuvettes
About This Item
UNSPSC Code:
41121813
NACRES:
SB.32
material
Suprasil® quartz
description
4 polished windows
manufacturer/tradename
Hellma 114F-QS
parameter
200-2,500 nm spectral range
H × W × D
46 mm × 12.5 mm × 12.5 mm
chamber volume
1,400 μL
1400 μL
pathlength
10x4 mm
General description
with PTFE stopper
Packaging
Sold as packages containing a single cell (1EA = 1 cell)
Legal Information
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.