Skip to Content
Merck
All Photos(1)

Key Documents

T7684

Sigma-Aldrich

Transaminase, broad-range bacterial

recombinant, expressed in E. coli, lyophilized powder, ≥5 units/mg solid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

form

lyophilized powder

specific activity

≥5 units/mg solid

storage temp.

2-8°C

General description

Increased transaminase activity alters the rate at which precursors of protein are interchanged with carbohydrate intermediates.

Application

Transaminase has been used in a study to assess the effect of ischemic preconditioning on pathophysiologic alterations of hemodynamic, biochemical and inflammatory parameters observed during intra-abdominal hypertension. Transaminase levels have also been used in a study to investigate the use of hydrogen-1 MR spectroscopy for measurement and diagnosis of hepatic steatosis.

Unit Definition

One unit will produce 1.0 μmole of oxaloacetate from L-aspartate during the transamination of 2-ketoglutarate per min at pH 7.0 at 25 °C in the presence of pyridoxal phosphate.

Physical form

Lyophilized powder containing potassium phosphate buffer salts

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucocorticosteroids and transaminase activity. I. Increased activity of glutamicpyruvic transaminase in four conditions associated with gluconeogenesis.
F ROSEN et al.
The Journal of biological chemistry, 234(3), 476-480 (1959-03-01)
Patrick Georgoff et al.
AJR. American journal of roentgenology, 199(1), 2-7 (2012-06-27)
Hydrogen-1 MR spectroscopy ((1)H-MRS) is gaining acceptance as a noninvasive technique for assessment of hepatic steatosis, and the findings have been found to correlate closely with histopathologic grade. The aims of this study were to validate (1)H-MRS performed with a
Alexandra Avraamidou et al.
Langenbeck's archives of surgery, 397(8), 1333-1341 (2012-07-05)
Intra-abdominal hypertension (IAH) has several pathophysiologic implications on human organs and systems. The aim of this experimental study was to investigate whether ischemic preconditioning (IP), namely the application of IAH for a small period of time prior to establish pneumoperitoneum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service