T0813
Anti-Tumor Necrosis Factor-α antibody produced in goat
affinity isolated antibody, lyophilized powder
Synonym(s):
Anti-TNF-α
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
51111800
NACRES:
NA.41
Recommended Products
biological source
goat
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
human
technique(s)
immunohistochemistry: suitable
neutralization: suitable
western blot: suitable
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... TNF(7124)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Tumor Necrosis Factor-α (TNF-α) is a cytokine produced by macrophages in response to stimulus by LPS. It plays a major role in mediating inflammation, tissue injury, pathogenic shock, innate immunity, apoptosis and autoimmunity. The physiological effects of TNF-α are mediated by receptors of TNFR super family. The TNF/TNFR signaling axis plays a critical role in mediating either survival or apoptosis depending on the class of adaptor proteins recruited in the cell. Association of receptors with a death domain causes rapid activation of caspases and results in cell death. TNF/TNFR also results in the activation of downstream pathways involving MAPK, p38, JNK and NF-κB that play a major role in innate immunity, acute inflammatory responses and homeostasis. The TNF/TNFR axis is also important in the pathology of various autoimmune and inflammatory disorders including a variety of malignancies and Alzheimer′s disease. Anti-Tumor Necrosis Factor-α antibody is specific for human TNF-α. The antibody shows less than 10% cross reactivity with recombinant rabbit TNF-α and less than 5% cross reactivity with recombinant mouse TNF-α.
Specificity
The antibody neutralizes the biological activity of recombinant human TNF-α. The antibody shows <10% cross-reactivity with recombinant rat TNF-α and <5% cross-reactivity with recombinant mouse TNF-α.
Immunogen
recombinant human TNF-α.
Application
Anti-Tumor Necrosis Factor-α (TNF- α) antibody produced in goat has been used to study the release of TNF- α from the cocultured lung carcinoma cells A427:AML-193.
Anti-Tumor Necrosis Factor-α antibody may be used for immunoblotting at a working antibody concentration of 0.1 μg/ml. For immunohistochemistry a concentration range of 5-15 μg/ml is recommended. The antibody may be used for neutralization assay; the ND50 is 0.01-0.06 μg/mL in the presence of 1.5ng/mL Recombinant Human TNF-alpha.
Physical form
Lyophilized from 0.2 μm filtered solution in phosphate buffered saline containing 5% trehalose.
Preparation Note
The antibody is purified using TNF-α affinity chromatography.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Biochanin A extirpates the epithelial-mesenchymal transition in a human lung cancer.
Wang Y, et al.
Experimental and Therapeutic Medicine, 15(3), 2830-2836 (2018)
The TNF receptor superfamily of cellular and viral proteins: activation, costimulation, and death.
C A Smith et al.
Cell, 76(6), 959-962 (1994-03-25)
K A Papadakis et al.
Annual review of medicine, 51, 289-298 (2000-04-25)
Recent advances in the drug treatment of inflammatory bowel disease (IBD) have paralleled our understanding of the pathophysiology of ulcerative colitis and Crohn's disease. Several proinflammatory and immune-regulatory cytokines are upregulated in the mucosa of patients with IBD, and differences
A Ashkenazi et al.
Science (New York, N.Y.), 281(5381), 1305-1308 (1998-08-28)
Apoptosis is a cell suicide mechanism that enables metazoans to control cell number in tissues and to eliminate individual cells that threaten the animal's survival. Certain cells have unique sensors, termed death receptors, on their surface. Death receptors detect the
The TNF and TNF receptor superfamilies: integrating mammalian biology.
R M Locksley et al.
Cell, 104(4), 487-501 (2001-03-10)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service