Skip to Content
Merck
All Photos(1)

Documents

SRP2089

Sigma-Aldrich

C-myc, proto oncogene human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

MRTL, bHLHe39, c-Myc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~50.4 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

500 μg/mL

color

clear
colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MYC(4609)

General description

MYC oncogene is a global transcription factor. The gene is located on human chromosome 8q24.21. MYC oncogene is a cancer promoting gene.

Biochem/physiol Actions

C-myc oncogene has high proliferative capacity. Overexpression of this gene is associated with Burkitt lymphoma. C-myc oncogene is implicated in various malignant tumors, such as, leukemia, lymphoma and human solid tumor.
The universal deregulation of c-Myc gene expression in tumor cells suggests that this oncogene represents an attractive target for cancer therapeutic purposes. The c-Myc promoter integrates diverse mitogenic signaling cascades, which are constitutively activated in tumor cells, and translates them into expression of the c-Myc transcription factor, which promotes cell proliferation, growth, differentiation, and apoptosis by regulating the expression of numerous target genes. The structural and biochemical features of the MYC family (MYC, N-MYC, and L-MYC) mark them as direct regulators of gene expression. As basic helix-loop-helix leucine zipper proteins (bHLH-ZIP), the MYCs acquire the capacity to bind the DNA sequence CACGTG (E-box) when dimerized with MAX (another bHLH-ZIP, 4,5). A head-to-tail pair of MYC-MAX dimers may, in turn, form a heterotetramer capable of bridging distant E-boxes. Among the broadly distributed positive enforcers of MYC action that are often recruited to target genes are chromatin remodeling (SWI/SNF relatives) and modifying complexes (TRAPP/GCN5 and relatives); these complexes mobilize nucleosomes and acetylate histones and/or other targets to activate gene expression. MYC binds TBP along an auxiliary pathway to control gene expression. MAD and MNT generally oppose MYC action by enlisting histone deacetylase complexes. Besides acting at the level of chromatin, MYC may also operate at later stages of the transcription cycle, after pre-initiation complex formation. In addition to using generic chromatin complexes to up- or down-regulate transcription, the MYC network also conscripts individual factors to modify expression locally on an ad hoc basis. For example, YY1, AP2, MIZ1, SP1, BRCA1, and other proteins interact directly with MYC, and so may directly modify the output of the MYC network.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel synthetic 4-chlorobenzoyl berbamine inhibits c-Myc expression and induces apoptosis of diffuse large B cell lymphoma cells
Zhang L, et al.
Annals of Hematology, 97(12), 2353-2362 (2018)
C V Dang et al.
Experimental cell research, 253(1), 63-77 (1999-12-02)
The c-myc gene and the expression of the c-Myc protein are frequently altered in human cancers. The c-myc gene encodes the transcription factor c-Myc, which heterodimerizes with a partner protein, termed Max, to regulate gene expression. Max also heterodimerizes with
Low expression of c-Myc protein predicts poor outcomes in patients with hepatocellular carcinoma after resection
Ji F, et al.
BMC Cancer, 18(1), 460-460 (2018)
Consistent MYC and FLT4 gene amplification in radiation-induced angiosarcoma but not in other radiation-associated atypical vascular lesions
Guo T, et al.
Genes Chromosomes Cancer, 50(1), 25-33 (2011)
Deconstructing myc.
R N Eisenman
Genes & development, 15(16), 2023-2030 (2001-08-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service