Skip to Content
Merck
All Photos(1)

Key Documents

SML2238

Sigma-Aldrich

NAV-2729

≥98% (HPLC)

Synonym(s):

3-(4-Chlorophenyl)-5-(4-nitrophenyl)-2-(phenylmethyl)-pyrazolo[1,5-a]pyrimidin-7(4H)-one, NAV 2729, NAV2729

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H17ClN4O3
CAS Number:
Molecular Weight:
456.88
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C=C(C2=CC=C([N+]([O-])=O)C=C2)NC3=C(C4=CC=C(Cl)C=C4)C(CC5=CC=CC=C5)=NN31

Biochem/physiol Actions

NAV-2729 (IC50 of 1.5 μM) has a therapeutic benefit in reducing the adverse effect of diabetic retinopathy in animal models.
Originally characterized as a non-nucleotide-competitive and reversible ARF6-selective inhibitor (IC50 = 1.4 μM without GEF and 2.4 μM with 100 nM ARNO or BRAG2/GEP100) that targets ARF6 GEF-binding region, NAV-2729 prevents GEF-dependent ARF1 & ARF6 activity (% inhibition of BRAG2Sec7PH-stimulated GTPase activity/[NAV-2729] = 50% Δ17Arf1/10 μM and 15% Δ13Arf6/25 μM) with higher potency against BRAG2- than ARNO-dependent ARF1 activity (64% vs. 20% Δ17Arf1 inhibition at 25 μM in the presence of respective GEF sec7 domain). NAV-2729 treatment effectively inhibits G-alpha-q downstream signaling pathways and anchorage-independent colony growth of Mel92.1 & Mel202 melanoma cells in vitro (10 μM) as well as uveal melanoma tumor establishment in Mel202 xenograft mice in vivo (30 mg/kg/day i.p.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Benabdi et al.
Biochemistry, 56(38), 5125-5133 (2017-09-01)
Arf GTPases and their guanine nucleotide exchange factors (ArfGEFs) are major regulators of membrane traffic and organelle structure in cells. They are associated with a variety of diseases and are thus attractive therapeutic targets for inhibition by small molecules. Several
Jae Hyuk Yoo et al.
Cancer cell, 29(6), 889-904 (2016-06-07)
Activating mutations in Gαq proteins, which form the α subunit of certain heterotrimeric G proteins, drive uveal melanoma oncogenesis by triggering multiple downstream signaling pathways, including PLC/PKC, Rho/Rac, and YAP. Here we show that the small GTPase ARF6 acts as
Weiquan Zhu et al.
The Journal of clinical investigation, 127(12), 4569-4582 (2017-10-24)
The devastating sequelae of diabetes mellitus include microvascular permeability, which results in retinopathy. Despite clinical and scientific advances, there remains a need for new approaches to treat retinopathy. Here, we have presented a possible treatment strategy, whereby targeting the small
Yohei Yamauchi et al.
Advances in biological regulation, 63, 115-121 (2016-10-26)
The Small GTPase ADP-ribosylation factor 6 (Arf6) functions as the molecular switch in cellular signaling pathways by cycling between GDP-bound inactive and GTP-bound active form, which is precisely regulated by two regulators, guanine nucleotide exchange factors (GEFs) and GTPase-activating proteins
Teclegiorgis Gebremariam et al.
Antimicrobial agents and chemotherapy, 64(8) (2020-05-13)
The rise in multidrug-resistant (MDR) organisms portends a serious global threat to the health care system with nearly untreatable infectious diseases, including pneumonia and its often fatal sequelae, acute respiratory distress syndrome (ARDS) and sepsis. Gram-negative bacteria (GNB), including Acinetobacter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service