Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700142

Sigma-Aldrich

Monoclonal Anti-CD80-FITC antibody produced in mouse

clone MEM-233, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-B7-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-233, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD80(941)

General description

Cluster of differentiation 80 (CD80), also called as lymphocyte activation antigen 7-1, is encoded by the gene mapped to human chromosome 3q13.33. The encoded protein is mainly expressed on antigen presenting cells such as, neutrophils, macrophages and dendritic cells.
The antibody MEM-233 reacts with CD80 (B7-1), a 60 kDa single chain type I glycoprotein of immunoglobulin supergene family, expressed on professional antigen-presenting cells, such as dendritic cells, macrophages or activated B lymphocytes.

Immunogen

Extracellular domain of human CD80 fused to human IgG1(Fc)

Application

The reagent is designed for Flow Cytometry analysis of human blood cells using 20 μL reagent / 100 μL of whole blood or 1e6 cells in a suspension. The content of a vial (2 mL) is sufficient for 100 tests.

Biochem/physiol Actions

Cluster of differentiation 80 (CD80) interacts with its ligand, co-stimulator ligand CD28 or with cytotoxic T-lymphocyte protein 4 (CTLA-4) and facilitates the regulation of CD4+ and CD8+ T cell activity. Elevated urinary CD80 excretion leads to Fabry disease. CD80, which is expressed on surface of dendritic cells acts as a key co-stimulatory molecule and induces second signal for the proper stimulation of cancer antigen-specific naive T cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association Analyses Identify Three Susceptibility Loci for Vitiligo in the Chinese Han Population
Tang XF
The Journal of Investigative Dermatology, 133, 403-410 (2013)
Increased urinary CD80 excretion and podocyturia in Fabry disease
Trimarchi H
Journal of Translational Medicine, 14 (2016)
Raquel Gil-Edo et al.
Pharmaceuticals (Basel, Switzerland), 16(6) (2023-06-28)
This work focuses on the development of thirteen benzylethylenearyl ureas and one carbamate. After the synthesis and purification of the compounds, we studied their antiproliferative action on cell lines, such as HEK-293, and cancer ones, such as HT-29, MCF-7 or
Prediction of Radix Astragali Immunomodulatory Effect of CD80 Expression from Chromatograms by Quantitative Pattern-Activity Relationship.
Ng MC
BioMed Research International (2017)
Exploring BenzylethoxyAryl Urea Scaffolds for Multitarget Immunomodulation Therapies.
Gil-Edo, et al.
International Journal of Molecular Sciences, 24 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service