Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500367

Sigma-Aldrich

Anti-Collagen III antibody produced in rabbit

affinity isolated antibody

Synonym(s):

COL3A1, Collagen α-1(III) chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 138 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL3A1(1281)

Related Categories

General description

Anti-Collagen III Antibody detects endogenous levels of total Collagen III protein.
Collagen III is also termed as collagen type III α1 (COL3A1). It is a fibrillar collagen present in extensible connective tissues. COL3A1 spans around 44 kb in length and has 52 exons. It is expressed in blood vessels and hollow organs. Collagen III gene is located on human chromosome 2q32.2.

Immunogen

The antiserum was produced against synthesized peptide derived from human Collagen III.

Immunogen Range: 81-130

Application

Anti-Collagen III antibody has been used in immunohistochemistry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Collagens usually help in the development of fibrillar and microfibrillar networks of the extracellular matrix, basement membranes and structures of the extracellular matrix. Mutations in the COL3A1 gene results in vascular ehlers-danlos syndrome (vEDS). COL3A1 controls the expression and modulation of collagen.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic variability in the extracellular matrix as a determinant of cardiovascular risk: association of type III collagen COL3A1 polymorphisms with coronary artery disease.
Muckian C, et al.
Blood, 100(4), 1220-1223 (2002)
The genetics and genomics of thoracic aortic disease.
Pomianowski P and Elefteriades JA
Annals of cardiothoracic surgery, 2(3), 271?279-271?279 (2013)
A new candidate substrate for cell-matrix adhesion study: the acellular human amniotic matrix.
Guo Q, et al.
BioMed Research International, 2012 (2012)
Transcriptome analysis of skin fibroblasts with dominant negative COL3A1 mutations provides molecular insights into the etiopathology of vascular Ehlers-Danlos syndrome.
Chiarelli N, et al.
PLoS ONE, 13(1), e0191220-e0191220 (2018)
miR-29a/b enhances cell migration and invasion in nasopharyngeal carcinoma progression by regulating SPARC and COL3A1 gene expression.
Qiu F, et al.
PLoS ONE, 10(3), e0120969-e0120969 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service