Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500065

Sigma-Aldrich

Anti-p21 CIP1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CAP20, CDK-interacting protein 1, CDKN1, CDKN1A, CDN1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 17 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDKN1A(1026)

General description

Anti-p21 CIP1 Antibody detects endogenous levels of total p21 CIP1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human p21 Cip1.

Immunogen Range: 111-160

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuqi Wu et al.
Molecular medicine reports, 19(5), 4433-4440 (2019-04-04)
MicroRNAs (miRNAs) are post‑transcriptional regulators that mediate the initiation and progression of human cancer. Growing evidence suggests that deregulation of miRNA expression levels underlies chemo‑resistance. To investigate whether miRNA‑302a (miR‑302a) is involved in mediating chemo‑resistance to paclitaxel in prostate cancer
T Feng et al.
Oncogenesis, 6(1), e294-e294 (2017-01-31)
TGF-β signaling in liver cells has variant roles in the dynamics of liver diseases, including hepatocellular carcinoma (HCC). We previously found a correlation of high levels of the important endogenous negative TGF-β signaling regulator SMAD7 with better clinical outcome in
Guijuan Liu et al.
Molecular medicine reports, 12(2), 2225-2230 (2015-04-16)
Cell proliferation is a major underlying cause of mortality amongst patients with oral squamous cell carcinoma (OSCC); however, the underlying mechanisms have remained to be elucidated. Acylglycerol kinase (AGK) is a multisubstrate lipid kinase, which is known to be associated
Jiantao Zheng et al.
Medical science monitor : international medical journal of experimental and clinical research, 22, 4431-4437 (2016-11-20)
BACKGROUND MicroRNAs have been reported to play significant roles in pathogenesis of colorectal cancer (CRC). In the present study, we aimed to investigate the functional role of microRNA-455-3p (miR-455-3p) in CRC, as well as its underlying mechanisms. MATERIAL AND METHODS
Han-Qing Zhu et al.
Cancer medicine, 8(11), 5313-5326 (2019-07-25)
Oridonin could induce NB (neuroblastoma) cells growth inhibition by inducing apoptosis and cell cycle arrest, and the molecular mechanisms behind the effects deserve to be further explored. Here, oridonin was confirmed to cause the reactivation of p53 (cellular tumor antigen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service