Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300627

Sigma-Aldrich

Anti-CCR7 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BLR2, Anti-CD197, Anti-CDw197, Anti-CMKBR7, Anti-EBI1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~43 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(T-D-D-Y-I)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCR7(1236)

Related Categories

General description

Anti-CCR7 antibody detects endogenous level of total CCR7(CD197) protein. CCR7 (C-C motif chemokine receptor 7) is localized to T cells, natural killer and dendritic cells. The CCR7 gene is mapped to human chromosome 17q21.2.

Immunogen

Peptide sequence around aa. 29-33 (T-D-D-Y-I), according to the protein NP_001829.1

Biochem/physiol Actions

CCR7 (C-C motif chemokine receptor 7) regulates lymphocyte cell trafficking and lymph node distribution. CCR7′s interaction with CCL21 promotes metastasis of cancer via lymph node. CCR7 is known to be associated with breast cancer, melanoma, colorectal cancer, non-small cell lung cancer, and chronic lymphocytic leukemia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Receptor for the MIP-3-beta chemokine. Probable mediator of EBV effects on B-lymphocytes or of normal lymphocyte functions.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of CXCR4, CCR7, VEGF-C and VEGF-D expression with lymph node metastasis in patients with cervical cancer.
Dai Y, et al.
Eur. J. Obstet. Gyn. Rep. Biol., 214, 178-183 (2017)
Tumor hypoxia modulates podoplanin/CCL21 interactions in CCR7+ NK cell recruitment and CCR7+ tumor cell mobilization.
Tejchman A, et al.
Oncotarget, 8(19), 31876-31876 (2017)
Confirmation of novel type 1 diabetes risk loci in families.
Cooper J D, et al.
Diabetologia, 55(4), 996-1000 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service