Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200343

Sigma-Aldrich

Anti-MLL2 antibody, Mouse monoclonal

clone MLL2-A, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-MLL2 antibody produced in mouse, Anti-AAD10, Anti-ALL1-related protein, Anti-ALR, Anti-CAGL114, Anti-Histone-lysine N-methyltransferase, Anti-KMS, Anti-KMT2B, Anti-KMT2D, Anti-Kabuki make-up syndrome, Anti-Kabuki mental retardation syndrome, Anti-MLL4, Anti-Myeloid/lymphoid or mixed-lineage leukemia 2, Anti-TNRC21

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

MLL2-A, monoclonal

form

buffered aqueous solution

mol wt

antigen ~17 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: 0.2-0.5 μg/mL using using human MLL2 partial fusion protein (immunogen)

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MLL2(8085)

Looking for similar products? Visit Product Comparison Guide

General description

Myeloid/lymphoid or mixed-lineage leukemia 2 (MLL2) belongs to the SET family of proteins. The SET domain of MLL2 confers strong histone 3 lysine 4 methyltransferase activity. MLL2 is part of a large protein complex called activating signal cointegrator-2 complex (ASCOM).

Immunogen

Fusion protein corresponding to the C-terminus of human MLL2. The corresponding sequence is identical in mouse and rat MLL2 and shares 85% homology with MLL3.

Application

Monoclonal Anti-MLL2 antibody produced in mouse has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

MLL2 acts as an epigenetic transcriptional activator during growth and development. It functions as a transcriptional regulator of the beta-globin and estrogen receptor genes. Mutations in this gene are linked to Kabuki syndrome.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PLK1 signaling in breast cancer cells cooperates with estrogen receptor-dependent gene transcription
Wierer M, et al.
Cell Reports, 3(6), 2021-2032 (2013)
ASCOM controls farnesoid X receptor transactivation through its associated histone H3 lysine 4 methyltransferase activity
Kim DH, et al.
Molecular Endocrinology, 23(10), 1556-1562 (2009)
Exome sequencing identifies MLL2 mutations as a cause of Kabuki syndrome
Ng SB, et al.
Nature Genetics, 42(9), 790-790 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service