Skip to Content
Merck
All Photos(1)

Key Documents

SAB3701172

Sigma-Aldrich

Anti-Mouse IgG1 (γ-chain specific)-Alkaline Phosphatase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

ALP, Alk Phos

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. It consists of a γ heavy chain in the constant (C) region. The monomeric 150kDa structure of IgG constitutes two identical heavy chains and two identical light chains with molecular weight of 50kDa and 25kDa, respectively. The primary structure of this antibody also contains disulfide bonds involved in linking the two heavy chains, linking the heavy and light chains and resides inside the chains. Maternal IgG is the only antibody transported across the placenta to the fetus. It passively immunizes the infants. IgG is further subdivided into four classes namely, IgG1, IgG2, IgG3, and IgG4 with different heavy chains, named γ1, γ2, γ3, and γ4, respectively. IgG1 is induced due to antibody responses to membrane proteins and soluble protein antigens. It is the most abundant subclass of IgG.

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using antigens coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Alkaline Phosphatase (calf intestine) and Anti-Rabbit Serum. Specificity was confirmed by ELISA at less than 1% cross reactivity against other Mouse heavy or light chain isotypes.

Immunogen

Mouse IgG1 heavy chain

Application

Anti-Mouse IgG1 (γ-chain specific)-Peroxidase antibody produced in rabbit has been used in ELISA.

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0 with 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of vaccinal effectiveness of preparations containing membrane antigens of Leishmania (L.) amazonensis in experimental cutaneous leishmaniasis model
Jo?o G.Ribeiro
International Immunopharmacology, 47, 227-230 (2017)
João G Ribeiro et al.
International immunopharmacology, 47, 227-230 (2017-04-24)
American tegumentary leishmaniasis (ATL) is considered a neglected disease, for which an effective vaccine or an efficient diagnosis is not yet available and whose chemotherapeutic arsenal is threatened by the emergence of resistance by etiological agents such as Leishmania amazonensis.
Gestur Vidarsson et al.
Frontiers in immunology, 5, 520-520 (2014-11-05)
Of the five immunoglobulin isotypes, immunoglobulin G (IgG) is most abundant in human serum. The four subclasses, IgG1, IgG2, IgG3, and IgG4, which are highly conserved, differ in their constant region, particularly in their hinges and upper CH2 domains. These
Human placental Fc receptors and the transmission of antibodies from mother to fetus.
Simister NE and Story CM
Journal of Reproductive Immunology, 37(1), 1-23 (1997)
Antibody structure, instability, and formulation.
Wang W
Journal of Pharmaceutical Sciences, 96(1), 1-26 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service