Skip to Content
Merck
All Photos(2)

Key Documents

SAB1409246

Sigma-Aldrich

Monoclonal Anti-IL8 antibody produced in mouse

clone 4E3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CXCL8, GCP-1, GCP1, LECT, LUCT, LYNAP, MDNCF, MONAP, NAF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4E3, monoclonal

form

buffered aqueous solution

mol wt

antigen 8.69 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL8(3576)

Related Categories

General description

Interleukin 8 (IL-8) is an 8& kDa, 72 amino acid residue containing cytokine that belongs to C-X-C chemokine family. The IL-8 gene is mapped to human chromosomes 4q13.3. IL-8 chemokine is secreted by several cell types. The IL-8 gene is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection.

Immunogen

IL8 (AAH13615, 21 a.a. ~ 99 a.a) full length recombinant protein.

Sequence
EGAVLPRSAKELRCQCIKTYSKPFHPKFIKELRVIESGPHCANTEIIVKLSDGRELCLDPKENWVQRVVEKFLKRAENS

Biochem/physiol Actions

Elevated expression of IL-8 gene is observed in a number of cancers including chronic spontaneous urticaria and breast cancer, thus serves as a prognostic marker for the same. IL-8 regulates the metastasis and angiogenesis processes during cancer development. IL-8 is responsible for epithelial–mesenchymal transition which plays an important role in providing cancer cells the mobility to invade other neighbouring cells, thereby contributing to metastasis.
Interleukin-8 (IL-8) chemoattracts and activates neutrophils. It is involved in guiding neutrophils and oligodendrocytes to sites of infection. The protein also stimulates angiogenesis and tumor growth. It is also associated with inflammatory diseases.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification of a human monocyte-derived neutrophil chemotactic factor that has peptide sequence similarity to other host defense cytokines.
Yoshimura T, et al.
Proceedings of the National Academy of Sciences of the USA, 84(24), 9233-9237 (1987)
Interleukin 8: cells of origin in inflammatory bowel disease.
Grimm M C, et al.
Gut, 38(1), 90-98 (1996)
Elevated plasma il-8 concentration is related to severity of systemic inflammation in chronic spontaneous urticaria.
Kasperska-Zajac A, et al.
Journal of Biological Regulators and Homeostatic Agents, 31(4), 957-961 (2017)
Sohlh2 suppresses epithelial to mesenchymal transition in breast cancer via downregulation of IL-8.
Ji S, et al.
Oncotarget, 7(31), 49411-49411 (2016)
The chemokine and chemokine receptor superfamilies and their molecular evolution.
Zlotnik A
Genome Biology, 7(12), 243-243 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service