Skip to Content
Merck
All Photos(1)

Key Documents

S9947

Sigma-Aldrich

Monoclonal Anti-Sonic Hedgehog Peptide (C-terminal) antibody produced in rat

clone 55626, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-Dsh, Anti-Hhg1, Anti-Hx, Anti-Shh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

55626, monoclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

western blot: 1-2 μg/mL

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Shh(20423)

Immunogen

purified, E. coli-derived recombinant mouse 6× histidine-tagged sonic hedgehog C-terminal peptide (aa 199-437).

Biochem/physiol Actions

Shh protein is secreted by Purkinje cells in the cerebellum with both autocrine and paracrine functions. The Shh signaling is mediated by the interaction of Shh with transmembrane proteins, Patched and Smoothened. The Shh pathway cross-talks Notch and Wnt pathways and acts via downstream targets such as cyclins, gamma catenin, myc and other transcription factors. The Hh family members have multiple roles as morphogens, cell survival factors, setting anterior-posterior axis and digit formation during development of limb, and axon guidance factors during the development of brain and nervous system.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aileen Rivell et al.
Biology open, 8(3) (2019-03-07)
Beyond its role in patterning the neural tube during embryogenesis, additional functions of Sonic hedgehog (Shh) in post-embryonic and mature brains have been coming into focus. However, the question of the abundance of endogenous Shh - the ligand of the
Alessia Omenetti et al.
American journal of physiology. Gastrointestinal and liver physiology, 294(3), G595-G598 (2008-01-26)
Hedgehog (Hh) signaling modulates tissue remodeling by controlling the fate of Hh-responsive cells. Healthy adult livers exhibit little Hh activity. However, cells involved in adult liver repair, including myofibroblasts and progenitors, are capable of producing and responding to Hh ligands.
Fergal C Kelleher
Carcinogenesis, 32(4), 445-451 (2010-12-28)
To conduct a systematic review of the role that the hedgehog signaling pathway has in pancreatic cancer tumorigenesis. PubMed search (2000-2010) and literature based references. Firstly, in 2009 a genetic analysis of pancreatic cancers found that a core set of
Shohreh F Farzan et al.
American journal of physiology. Gastrointestinal and liver physiology, 294(4), G844-G849 (2008-02-02)
The Hedgehog (Hh) family of secreted proteins is necessary for aspects of the development and maintenance of the gastrointestinal tract. Hh is thought to function as a morphogen, a mitogen, a cell survival factor, and an axon guidance factor. Given

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service