Skip to Content
Merck
All Photos(4)

Key Documents

S8884

Sigma-Aldrich

Safranin O

Dye content, ≥80%, certified by the Biological Stain Commission, powder

Synonym(s):

Basic Red 2, Cotton Red, Gossypimine, Safranin T, Safranin Y or A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19ClN4
CAS Number:
Molecular Weight:
350.84
Colour Index Number:
50240
Beilstein:
3924099
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Safranin O, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

form

powder

composition

Dye content, ≥80%

color

green to very dark green

solubility

H2O: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].Cc1cc2nc3cc(C)c(N)cc3[n+](-c4ccccc4)c2cc1N

InChI

1S/C20H18N4.ClH/c1-12-8-17-19(10-15(12)21)24(14-6-4-3-5-7-14)20-11-16(22)13(2)9-18(20)23-17;/h3-11H,1-2H3,(H3,21,22);1H

InChI key

OARRHUQTFTUEOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Safranin O is a metachromatic, cationic dye. It is used as a counterstain in Gram staining. The stain colors Gram-negative bacteria pink to red and has no effect on Gram-positive bacteria. It is also used to assess glycosaminoglycans (GAGs) in cartilage. The dye imparts an intense red to GAGs. It easily binds to polyanions.

Application

Safranin O has been used in the staining of extraocular muscle mesenchymal stem cells (EOM-MSC) for the detection of cartilage, mucin and mast cell granules It has also been used as a mediator dye in cytochrome P450 spectral assay.
Safranin O has been used to stain matrix proteoglycans to study cartilaginous tissue structural integrity in mice. It has also been used to stain chondrogenically differentiated human bone marrow stromal cells.

Suitability

Certified as a stain for chromosomes in the Flemming triple stain; to stain gram negative bacteria in the Gram stain procedure; Safranine-fast green FCF stain for plant cells.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Novel markers of osteogenic and adipogenic differentiation of human bone marrow stromal cells identified using a quantitative proteomics approach
CeciliaGraneli
Stem Cell Research (2014)
Perturbations in the HDL metabolic pathway predispose to the development of osteoarthritis in mice following long-term exposure to western-type diet
I.-E.Triantaphyllidou
Osteoarthritis and Cartilage / OARS, Osteoarthritis Research Society (2013)
Isolation of Multipotent Mesenchymal Stem Cells from Human Extraocular Muscle Tissue
Sharma A, et al.
Bio-protocol, 9(4) (2016)
Paul A. Banaszkiewicz
Classic Papers in Orthopaedics (2014)
Maria Dannessa Delost
Introduction to Diagnostic Microbiology for the Laboratory Sciences (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service