Key Documents
S6815
Sigma-Aldrich
Nunc® Lab-Tek® II - CC2 Chamber Slide system
8 wells, glass slide, 0.7 cm2/well, sterile
Synonym(s):
cell culture chamber, chmaber slides, microscope slides, microscopy slides
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
Pricing and availability is not currently available.
material
colorless
polystyrene chamber
sterility
sterile
sterile
feature
CE marked
manufacturer/tradename
Nunc 154941
packaging
pkg of carton of 16
well area
0.7 cm2
wells
8
working volume
0.2-0.5 mL
suitability
suitable for (cell culture; optimal for culturing fastidious cells (e.g. neurons))
General description
Nunc® Lab-Tek® II - CC2 Chamber Slide System
  • Chemically coated growth surface on glass slide that mimics polylysine
  • Provides binding sites optimal for fastidious cells (e.g. neurons)
  • Growth surface remains stable without refrigeration
  • Light blue frosted writing area
  • Slide seperator included in each pack
  • CE marked
Legal Information
CC2 is a trademark of Thermo Fisher Scientific or its subsidiaries
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Christian Hinderer et al.
Human molecular genetics, 26(19), 3837-3849 (2017-09-22)
The mucopolysaccharidoses (MPS) are rare genetic disorders marked by severe somatic and neurological symptoms. Development of treatments for the neurological manifestations of MPS has been hindered by the lack of objective measures of central nervous system disease burden. Identification of
A Sundqvist et al.
Oncogene, 32(31), 3606-3615 (2012-08-29)
Deregulation of the transforming growth factor β (TGFβ) signal transduction cascade is functionally linked to cancer. In early phases, TGFβ acts as a tumor suppressor by inhibiting tumor cell proliferation, whereas in late phases, it can act as a tumor
Questions
1–2 of 2 Questions  
  1. Can growth chamber be removed when culturing is complete?
    1 answer
    1. Yes, the chambers can be removed after culture. Lab-Tek II slides use a hydrophobic acrylic adhesive that remains on the slide after chamber removal but, due to its very low profile, will not interfere with coverslips.
      Helpful?
  2. What are the characteristics of this product? I mean, is it just one chamber, or are there trays with multiple chambers?
    1 answer
    1. This product features a glass slide as the base, supporting a polystyrene chamber divided into 8 separate wells. The glass surface is tissue culture treated. The chambers are leak-proof and sealed at the base to prevent cross-contamination. One package includes 16 chambered slides. No additional accessories are included. Please review the image below:
      Helpful?