Skip to Content
Merck
All Photos(4)

Key Documents

P4903

Sigma-Aldrich

poly(C)

Synonym(s):

Polycytidylic acid potassium salt, Poly(C) potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.55

form

powder

Quality Level

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Prepared from CDP using polynucleotide phosphorylase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adalena V Nanni et al.
RNA biology, 15(9), 1181-1191 (2018-09-04)
Epstein-Barr virus (EBV) expresses an abundant nuclear noncoding RNA called EBER2, which interacts with and acts as a guide RNA for the host transcription factor PAX5. This ribonucleoprotein complex localizes to the terminal repeat (TR) regions of the EBV genome
Kang Xu et al.
Developmental and comparative immunology, 99, 103410-103410 (2019-06-09)
As a member of NDR protein kinase family and a novel protein kinase of Hippo signal pathway, Serine/threonine kinase 38 (STK38) plays a very significant role in the innate immune. In mammals, STK38 performs its function by combining with GSK3β.
Dongrong Yi et al.
Frontiers in microbiology, 11, 1440-1440 (2020-07-17)
All RNA viruses encode the RNA-dependent RNA polymerase (RdRp) which replicates and transcribes viral RNA. This essential viral enzyme does not exist in mammalian cells, thus presents a main target for the development of antiviral drugs with potential pan-antiviral activity.
Jordina Guillén-Boixet et al.
Cell, 181(2), 346-361 (2020-04-18)
Stressed cells shut down translation, release mRNA molecules from polysomes, and form stress granules (SGs) via a network of interactions that involve G3BP. Here we focus on the mechanistic underpinnings of SG assembly. We show that, under non-stress conditions, G3BP
Christina Kwong et al.
Oncoimmunology, 2(1), e22978-e22978 (2013-03-14)
The vacuolar ATPase (V-ATPase) plays an important role in tumor progression and metastases. A novel peptide from the a2 isoform of V-ATPase called a2NTD has been shown to exert an immunoregulatory role in the tumor microenvironment by controlling the maturation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service