Skip to Content
Merck
All Photos(1)

Key Documents

P3869

Supelco

Phosphorus standard solution

0.65 mM (phosphorous)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

Quality Level

100
200

concentration

0.65 mM (phosphorous)

storage temp.

2-8°C

Application

  • Antibacterial mode of action of 1,8-dihydroxy-anthraquinone from Porphyra haitanensis against Staphylococcus aureus.: This research details the antibacterial properties of compounds extracted from Porphyra haitanensis, using phosphorus standards in the evaluation process to ensure precise measurement and effective concentration assessment, crucial for developing biopharmaceutical applications (Wei Y et al., 2015).

Components

Phosphorus (as KH2PO4), 20 μg/mL in 0.05 N HCl.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cyntia Ayumi Yokota Harayashiki et al.
Marine pollution bulletin, 164, 112075-112075 (2021-01-31)
Studies have shown that shell morphology and enzymatic activities in mollusks are affected by contaminants exposure. However, the correlation between enzymatic activities and the biomineralization process are not fully understood. The present study used a transplant bioassay and field sampling
Ganesan Velmurugan et al.
Genome biology, 18(1), 8-8 (2017-01-25)
Organophosphates are the most frequently and largely applied insecticide in the world due to their biodegradable nature. Gut microbes were shown to degrade organophosphates and cause intestinal dysfunction. The diabetogenic nature of organophosphates was recently reported but the underlying molecular
Argiris Papathanasopoulos et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 29(9), 1327-1335 (2011-03-25)
An unexpected side effect of some classes of anticoagulants has been osteoporosis which may be, at least in part, related to deranged mesenchymal stem cell (MSC) function. The aim of the present study was to compare the effect of fondaparinux

Protocols

To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.

To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.

To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.

To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service