O6001
Octyl-Sepharose™ CL-4B
Synonym(s):
Octyl–Agarose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
extent of labeling
~40 μmol per mL
matrix
agarose, 4% cross-linked
matrix activation
epichlorohydrin
matrix attachment
ether
matrix spacer
3 atoms
particle size
45-165 μm
capacity
3-5 mg/mL binding capacity (β-lactoglobulin)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
O6001-200ML′s updated product number is GE17-0790-01
Application
Octyl Sepharose™ is used in hydrophobic interaction media, resins and separation media. Octyl Sepharose™ has been used to determine the population of Acinetobacter species in sewage treatment plants.
Physical form
Suspension in 24% ethanol
Legal Information
Sepharose is a trademark of Cytiva
related product
Product No.
Description
Pricing
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The Journal of biological chemistry, 271(48), 30347-30353 (1996-11-29)
Decorin, a member of a family of proteins with leucine-rich repeat motifs, is a widely distributed extracellular matrix proteoglycan that is thought to be responsible for the structure, tissue organization, and surface properties of fibrils. In mammals, decorin carries one
Journal of bacteriology, 190(7), 2379-2387 (2008-02-05)
Lipoteichoic acid (LTA) is an essential bacterial membrane polysaccharide (cell wall component) that is attached to the membrane via a lipid anchor. According to the currently accepted structure of pneumococcal LTA, the polysaccharide is comprised of several repeating units, each
Biochimica et biophysica acta, 523(2), 314-320 (1978-04-12)
1. Hydrophobic interaction chromatography on Octyl-Sepharose CL-4B is used as a new and simple method for the preparation of large amounts of beef-heart cytochrome c oxidase (ferrocytochrome c:oxygen oxidoreductase, EC 1.9.3.1). 2. The method involves only one cycle of (NH4)2SO4
The Biochemical journal, 282 ( Pt 3), 711-714 (1992-03-15)
Epoxide hydrolases catalysing the hydration of cis-9,10-epoxystearate into threo-9,10-dihydroxystearate have been detected in soybean (Glycine max) seedlings. The major activity was found in the cytosol, a minor fraction being strongly associated with microsomes. The soluble enzyme, which was purified to
Biochimica et biophysica acta, 1788(12), 2526-2540 (2009-10-06)
Gangliosides are glycolipids mainly present at the plasma membrane (PM). Antibodies to gangliosides have been associated with a wide range of neuropathy syndromes. Particularly, antibodies to GM1 ganglioside are present in patients with Guillain-Barré syndrome (GBS). We investigated the binding
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service