Skip to Content
Merck
All Photos(1)

Key Documents

O1516

Sigma-Aldrich

Oil Red O solution

0.5% in propylene glycol

Synonym(s):

Solvent Red 27, Sudan Red 5B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24N4O
CAS Number:
Molecular Weight:
408.49
UNSPSC Code:
12171500
NACRES:
NA.47

form

liquid

Quality Level

concentration

0.5% in propylene glycol

technique(s)

microbe id | staining: suitable

λmax

0.40-1.00 at 506-520 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

N(\N=C3\c4c(cccc4)C=CC\3=O)c1c(cc(c(c1)C)N=Nc2c(ccc(c2)C)C)C

InChI

1S/C26H24N4O/c1-16-9-10-17(2)22(13-16)27-28-23-14-19(4)24(15-18(23)3)29-30-26-21-8-6-5-7-20(21)11-12-25(26)31/h5-15,29H,1-4H3/b28-27?,30-26-

InChI key

HJAYEODIXUYVIC-LTFAAIOASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oil Red O stains and imparts red-orange color to lipids. It has also been shown to mark vacuoles, observed in acute lymphoblastic leukemia (ALL-L3) or Burkitt′s lymphoma/leukemia. Oil Red O staining is done on fresh samples, as alcohol fixation removes lipids.

Application

Oil red O is a lysochrome (fat-soluble dye) diazo dye used for staining of neutral triglycerides and lipids on frozen sections and some lipoproteins on paraffin sections. In histology, a supersaturated solution of oil red O in isopropanol may be used to stain fat in tissue.
Oil red O solution has been used to identify intracytoplasmatic lipid droplets on 3T3-L1 cell line adipocytes and to measure lipid content in Simpson-Golabi-Behmel syndrome (SGBS) preadipocytes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F

Flash Point(C)

103 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alyssa M Flores et al.
Nature nanotechnology, 15(2), 154-161 (2020-01-29)
Atherosclerosis is the process that underlies heart attack and stroke. A characteristic feature of the atherosclerotic plaque is the accumulation of apoptotic cells in the necrotic core. Prophagocytic antibody-based therapies are currently being explored to stimulate the phagocytic clearance of
Effect of Resveratrol on Mitochondrial Activity in Differentiated Mature Adipocytes.
Noriega-Gonzalez J E, et al.
International Journal of Morphology, 33(3) (2015)
LWW Doody's all reviewed collection, McClatchey: Clinical Laboratory Medicine
Clinical Laboratory Medicine (2002)
LipiD-QuanT: a novel method to quantify lipid accumulation in live cells.
Varinli H, et al.
Journal of Lipid Research, 56(11), 2206-2216 (2015)
Francesco Albanese
Canine and Feline Skin Cytology: A Comprehensive and Illustrated Guide to the Interpretation of Skin Lesions via Cytological Examination (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service