Skip to Content
Merck
All Photos(1)

Key Documents

L5667

Sigma-Aldrich

LB Agar Ampicillin-100, Plates

pre-poured LB agar plates with 100 μg/ml ampicillin

Synonym(s):

ampicillin agar plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

grade

for molecular biology

Quality Level

sterility

sterile; autoclaved

form

plates (10 cm)
plates (10cm, 20-25ml each)

availability

available only in USA

capacity

20-25 mL

storage temp.

2-8°C

suitability

selective for Escherichia coli
selective for coliforms

General description

Miller′s LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates in a low-salt formulation. The addition of agar provides a solid medium for microbial growth.

Application

LB Agar Ampicillin-100, Plates has been used to plate and incubate the mixture of thawed bacteria and plasmid solution for vimentin expression. It has also been used in the generation of the Trypanosoma brucei RNAi library.

Features and Benefits

Miller′s LB Agar plates provide:
  • Convenient, ready-to-use 10cm format
  • Standard formulation plus antibiotic

Components

15g/L Agar
10g/L Tryptone
10g/L NaCl
5g/L Yeast Extract
100μg/ml ampicillin

Other Notes

The plates must be used within three months of the manufacture date to ensure the antibiotics are fully active. The product may be shipped up until its expiration date but is generally shipped 2-4 weeks before expiration.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-scale RNAi screens for high-throughput phenotyping in bloodstream-form African trypanosomes
Glover L, et al.
Nature Protocols, 10(1), 106-106 (2015)
Viscoelastic properties of vimentin originate from nonequilibrium conformational changes
Block J, et al.
Science advances, 4(6) (2018)
Linfeng Xu et al.
Nature communications, 12(1), 6803-6803 (2021-11-25)
Enzymes are represented across a vast space of protein sequences and structural forms and have activities that far exceed the best chemical catalysts; however, engineering them to have novel or enhanced activity is limited by technologies for sensing product formation.
Viscoelastic properties of vimentin originate from nonequilibrium conformational changes
Block J, et al.
Science Advances, 4(6) (2018)
Daniel Del Valle Morales et al.
Bio-protocol, 10(20), e3791-e3791 (2021-03-05)
The 5' cap is a ubiquitous feature of eukaryotic mRNAs. It is added in the nucleus onto newly synthesized pre-mRNA, and in the cytoplasm onto mRNAs after decapping or endonuclease cleavage. Cytoplasmic recapping can occur after loss of the cap

Articles

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service