Skip to Content
Merck
All Photos(2)

Documents

L0506

Sigma-Aldrich

Arachidyl dodecanoate

≥99%

Synonym(s):

Arachidyl laurate, Dodecanoic acid eicosyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H64O2
CAS Number:
Molecular Weight:
480.85
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99%

lipid type

unsaturated FAs

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCC

InChI

1S/C32H64O2/c1-3-5-7-9-11-13-14-15-16-17-18-19-20-21-23-25-27-29-31-34-32(33)30-28-26-24-22-12-10-8-6-4-2/h3-31H2,1-2H3

InChI key

GTUGKPSGIYCKEQ-UHFFFAOYSA-N

Biochem/physiol Actions

Arachidyl dodecanoate (arachidyl laurate) is a laurate medium-chain saturated (C20:0) fatty acid ester compound that may be used as a reference in the analysis of wax esters of lauric acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Pérez-Camino et al.
Journal of chromatography. A, 983(1-2), 283-288 (2003-02-06)
A procedure for the simultaneous determination of long-chain aliphatic aldehydes, and aliphatic and triterpenic waxes in virgin olive oils is described. A fraction containing these compounds was isolated from the oil using solid-phase extraction on silica-gel cartridges. The fraction was
Li-Hsueh Wang et al.
PloS one, 8(8), e72486-e72486 (2013-09-07)
Lipids are a source of metabolic energy, as well as essential components of cellular membranes. Although they have been shown to be key players in the regulation of cell proliferation in various eukaryotes, including microalgae, their role in the cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service