Skip to Content
Merck
All Photos(1)

Key Documents

I1161

Sigma-Aldrich

ImmunoHistoMount

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

Application

ImmunoHistoMount is a permanent aqueous mounting medium designed for tissue sections and cell smears with peroxidase and alkaline phosphatase chromogens that cannot be dehydrated with organic solvents. This mounting medium preserves fast red, aminoethylycarbazole (AEC), BCIP/NBT, and BCIP/INT chromogens and is compatible with counterstains like hematoxylin and nuclear fast red (NFR). ImmunoHistoMount is also suitable for chromogens like DAB and DAB with nickel and cobalt. It is not compatible with routine H & E staining.

Linkage

ImmunoHistoMount is an aqueous-based mounting medium comparable to Crystal Mount

Legal Information

Crystal Mount is a trademark of Biomeda Corp.
ImmunoHistoMount is a trademark of ImmunoBioScience Corp.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seizure frequency correlates with loss of dentate gyrus GABAergic neurons in a mouse model of temporal lobe epilepsy.
Buckmaster P S, et al.
The Journal of Comparative Neurology, 525(11), 2592-2610 (2017)
The effect of moving point of contact stimulation on chondrocyte gene expression and localization in tissue engineered constructs.
Kaupp J A, et al.
Annals of Biomedical Engineering, 41(6), 1106-1119 (2013)
Therapeutic immunization and local low-dose tumor irradiation, a reinforcing combination.
Draghiciu O, et al.
International Journal of Cancer. Journal International Du Cancer, 134(4), 859-872 (2014)
Damon T Jacobs et al.
Disease models & mechanisms, 9(7), 789-798 (2016-08-03)
Primary cilia extend from the plasma membrane of most vertebrate cells and mediate signaling pathways. Ciliary dysfunction underlies ciliopathies, which are genetic syndromes that manifest multiple clinical features, including renal cystic disease and obesity. THM1 (also termed TTC21B or IFT139)
Fernanda Medina-Flores et al.
Brain, behavior, and immunity, 89, 118-132 (2020-06-03)
Sleep loss in the rat increases blood-brain barrier permeability to circulating molecules by disrupting interendothelial tight junctions. Despite the description of the ultrastructure of cerebral microvessels and the evidence of an apparent pericyte detachment from capillary wall in sleep restricted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service