Skip to Content
Merck
All Photos(2)

Key Documents

HPA014861

Sigma-Aldrich

Anti-SLC31A2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Copper transporter 2, Anti-Probable low affinity copper uptake protein 2, Anti-Solute carrier family 31 member 2, Anti-hCTR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

YEGIKVGKAKLLNQVLVNLPTSISQQTIAETDGDSAGSDSFPVGRTHHRWYLC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC31A2(1318)

General description

SLC31A2 (solute carrier family 31 (copper transporter), member 2) is a copper transporter, and shares high homology to copper transporter1 (CTR1). It is also called CTR2. It has a highly conserved second transmembrane domain, which also contains MXXXM motif. This protein resides in lysosomes and late endosomes.

Immunogen

Probable low affinity copper uptake protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The function of SLC31A2 (solute carrier family 31 (copper transporter), member 2) is not fully understood yet. It exists as multimers or forms complexes with CTR2 (copper transporter 2). It is responsible for the uptake of Cu+1 by cells. In cells where it localizes to plasma membrane, it elevates Cu influx. Thus, it is involved in copper homeostasis. It is responsible for the transport of platinum drugs such as, cisplatin. It is up-regulated in chemoresistant ovarian cancer cases, where it confers platinum resistance to cancer cells, and this is related to poor prognosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73062

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian G Blair et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 15(13), 4312-4321 (2009-06-11)
Copper transporter 2 (CTR2) is known to mediate the uptake of Cu(+1) by mammalian cells. Several other Cu transporters, including the influx transporter CTR1 and the two efflux transporters ATP7A and ATP7B, also regulate sensitivity to the platinum-containing drugs. We
Hiroyuki Yoshida et al.
Anticancer research, 33(4), 1409-1414 (2013-04-09)
Copper transporters (CTR) also regulate the cellular transport of platinum drugs, but their role in platinum resistance of ovarian cancer has not been elucidated. CTR expression in ovarian cancer tissues resected from patients treated by platinum-based chemotherapy was evaluated immunohistochemically.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service