Skip to Content
Merck
All Photos(6)

Key Documents

HPA002321

Sigma-Aldrich

Anti-MVP antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Lung resistance-related protein antibody produced in rabbit, Anti-Major vault protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VFEEVLDLVDAVILTEKTALHLRARRNFRDFRGVSRRTGEEWLVTVQDTEAHVPDVHEEVLGVVPITTLGPHNYCVILDPVGPDGKNQLGQKRVVKGEKSFFLQPGEQLEQGIQDVYVLS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MVP(9961)

Related Categories

General description

Major vault protein (MVP) is the major vault protein with molecular mass of 110kDa. It is involved in the nucleo-cytoplasmic transport. It encodes for multidrug resistance-associated protein and protein kinase C-β and may have impact on the drug resistance (doxorubicin, vincristine, etoposide, paclitaxel, and gramicidin D) in cells via transport process.

Immunogen

Major vault protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MVP (major vault protein) is primarily responsible for maintaining the vault structure. In addition to maintenance of vault sturture, MVP is also involved in wide range of cellular activities such as drug resistance, subcellular transport, cell differentiation, regulation of intracellular signaling cascades including the PI (phosphoinositide) 3-kinase and the MAP (mitogen activated protein) kinase pathway. It acts as a Src-SH2 binding protein in human stomach tissue. MVP translocate from the nucleus to the cytosol and perinuclear region by the stimulation of epidermal growth factor (EGF). Thus, it proves MVP has high impact on Src-mediated cell signalling cascades. It has also been reported that MVP can stimulate EGF-dependent transcriptional activation for cell survival signaling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86064

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kitazono et al.
Journal of the National Cancer Institute, 91(19), 1647-1653 (1999-10-08)
Lung resistance-related protein (LRP), the major vault protein in humans, is sometimes overexpressed in multidrug-resistant cells. Because cells transfected with the LRP gene did not express the multidrug-resistant phenotype, we investigated whether LRP is involved in multidrug resistance. SW-620 cells
Elisabeth Steiner et al.
Journal of cell science, 119(Pt 3), 459-469 (2006-01-19)
The major vault protein (MVP) is the main component of vaults, large ribonucleoprotein particles implicated in the regulation of cellular signaling cascades and multidrug resistance. Here, we identify MVP as an interferon gamma (IFN-gamma)-inducible protein. Treatment with IFN-gamma resulted in
Euikyung Kim et al.
The FEBS journal, 273(4), 793-804 (2006-01-31)
Vaults are highly conserved, ubiquitous ribonucleoprotein (RNP) particles with an unidentified function. For the three protein species (TEP1, VPARP, and MVP) and a small RNA that comprises vault, expression of the unique 100-kDa major vault protein (MVP) is sufficient to
Sivanagarani Kolli et al.
The Journal of biological chemistry, 279(28), 29374-29385 (2004-05-11)
The catalytic activity of the Src homology 2 (SH2) domain-containing tyrosine phosphatase, SHP-2, is required for virtually all of its signaling effects. Elucidating the molecular mechanisms of SHP-2 signaling, therefore, rests upon the identification of its target substrates. In this
G L Scheffer et al.
Nature medicine, 1(6), 578-582 (1995-06-01)
Multidrug-resistant cancer cells frequently overexpress the 110-kD LRP protein (originally named Lung Resistance-related Protein). LRP overexpression has been found to predict a poor response to chemotherapy in acute myeloid leukaemia and ovarian carcinoma. We describe the cloning and chromosome localization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service