Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5451-01

Capto Phenyl High Sub

Cytiva 17-5451-01, pack of 25 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

ligand

phenyl

packaging

pack of 25 mL

manufacturer/tradename

Cytiva 17-5451-01

matrix

highly cross-linked agarose

avg. part. size

75 μm (d50v)

cleaning in place

2-14

working range

3-13

capacity

27 mg binding capacity(27 mg BSA /ml (at QB10%))

suitability

suitable for bioprocess medium

General description

Useful for the intermediate stages of protein purification, Capto Phenyl (High Sub) resin combines high capacity, high flow rate, and low back pressure to reduce process cycle times.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service