EP5424702007
Eppendorf® Rotor FA-45-24-11
incl. lid, aerosol tight
All Photos(1)
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Recommended Products
feature
to hold 24 x 1.5/2.0 Ml tubes
incl. lid, aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5424702007
parameter
15,000 max. RCF (21,130 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5424/5424R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)