Skip to Content
MilliporeSigma

Skip To

D7909

Dicyclomine hydrochloride

≥99% (TLC), powder

Synonym(s):

2-(Diethylamino)ethyl 1-cyclohexylcyclohexane-1-carboxylate hydrochloride, [1,1-Bicyclohexyl]-1-carboxylic acid 2-(diethylamino)ethyl ester hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g

Available to ship TODAYfromMILWAUKEE

$51.40

About This Item

Empirical Formula (Hill Notation):
C19H35NO2 · HCl
CAS Number:
Molecular Weight:
345.95
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-671-1
MDL number:
Assay:
≥99% (TLC)
Form:
powder

$51.40


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (TLC)

form

powder

color

off-white

solubility

H2O: 50 mg/mL

SMILES string

CCN(CCOC(C1(C2CCCCC2)CCCCC1)=O)CC.[Cl]

InChI

1S/C19H35NO2.ClH/c1-3-20(4-2)15-16-22-18(21)19(13-9-6-10-14-19)17-11-7-5-8-12-17;/h17H,3-16H2,1-2H3;1H

InChI key

GUBNMFJOJGDCEL-UHFFFAOYSA-N

Biochem/physiol Actions

Competitive muscarinic acetylcholine receptor antagonist.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
O2881D75711191000
form

powder

form

powder

form

powder

form

-

assay

≥99% (TLC)

assay

≥98% (TLC)

assay

≥98% (NMR)

assay

-

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

-

solubility

H2O: 50 mg/mL

solubility

H2O: 50 mg/mL

solubility

H2O: ≥50 mg/mL

solubility

-

color

off-white

color

white

color

white to off-white

color

-

Gene Information

human ... CHRM1(1128), CHRM2(1129), CHRM3(1131), CHRM4(1132), CHRM5(1133)

Gene Information

human ... CHRM1(1128), CHRM2(1129), CHRM3(1131), CHRM4(1132), CHRM5(1133)

Gene Information

-

Gene Information

human ... CHRM1(1128), CHRM3(1131)


Still not finding the right product?

Explore all of our products under Dicyclomine hydrochloride


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters