Skip to Content
Merck
All Photos(1)

Key Documents

CLS356008

Corning® Fibronectin

from human(Natural), pack of 5 mg

Synonym(s):

Cell adhesion protein, Fibronectin protein, Fibronectin solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NB.35

biological source

human (Natural)

packaging

pack of 5 mg

manufacturer/tradename

Corning 356008

Looking for similar products? Visit Product Comparison Guide

General description

Corning Fibronectin, human, 5mg, is used as a thin coating on tissue-culture surfaces to promote attachment, spreading, and proliferation of a variety of cell types. The principal functions of fibronectin appear to be in cellular migration during wound healing and development, regulation of cell growth and differentiation, and haemostasis/thrombosis.
Quality
Purity = 90% by SDS-PAGE
Tested for ability to promote attachment and spreading of BHK-21 cells
Found negative for bacteria, fungi, and mycoplasma

Source
Human plasma. Source material tested for hepatitis B antigen and HIV-1 antibody.

Molecular Weight
440 kD (unreduced form)

Preparation and Storage
Stable for three months at 2° to 8°C (lyophilized) or two weeks at -20°C (solubilized)

Handling
Recommended concentration - 1-5 μg/cm² of growth surface for coating or at 5 μg/ml as a media additive

Formulation
Lyophilized from CAPS buffer containing NaCl and CaCl2, pH 11.0

Legal Information

Corning is a registered trademark of Corning, Inc.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service