Skip to Content
Merck

Skip To

C6079

Collagenase + protease inhibitor

2-5 FALGPA units/mg solid, ≥800 CDU/mg solid

Synonym(s):

Collagenase preparation, Protease inhibitor solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

EC Number:
NACRES:
NA.54
UNSPSC Code:
12352204
Form:
solid
Solubility:
water: soluble
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bacterial (Clostridium histolyticum)

Quality Segment

form

solid

specific activity

≥800 CDU/mg solid, 2-5 FALGPA units/mg solid

mol wt

68-130 kDa

solubility

water: soluble

shipped in

dry ice

storage temp.

−20°C

Gene Information

Application

Collagenase with protease inhibitor has been used in a study to assess the association of immune system gene polymorphisms with quantitative features. It has also been used in a study to investigate the quantitative structure-activity relationship study on Clostridium histolyticum collagenase inhibitors. The enzyme from sigma has been used in the isolation of porcine pancreatic islets.[1] It has also been used in the isolation of pancreatic-infiltrating lymphocytes from mice.[2]

Biochem/physiol Actions

Effective release of cells from tissue requires the action of both collagenase enzymes and the neutral protease. Collagenase is activated by four grams of calcium (Ca2+) per mole of the enzyme. The culture filtrate is thought to contain at least 7 different proteases ranging in molecular weight from 68-130 kDa. The pH optimum is 6.3-8.8. The enzyme is typically used to digest the connective components in tissue samples to liberate individual cells. Collagenase treatment can cause some cells to die. Typically, concentrations varying from 0.1 to 5 mg/mL are used for digestion. The duration of reaction can vary from 15 minutes to several hours for satisfactory cell dissociation without causing too much cell death. Zn2+ is required for activity. This product is used if the collagenase does not require a significant protease activity.

Analysis Note

Also contains clostripain and non-specific neutral protease activities.

Other Notes

Selected lots of collagenase Type XI blended with protease inhibitor to limit the tryptic enzyme activities in pancreatic tissue. The formulation was developed through collaboration with several outside laboratories.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C2799C7926C2674
form

solid

form

powder

form

powder

form

lyophilized powder

biological source

bacterial (Clostridium histolyticum)

biological source

Clostridium histolyticum

biological source

Clostridium histolyticum

biological source

Clostridium histolyticum

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

solubility

water: soluble

solubility

-

solubility

-

solubility

-

shipped in

dry ice

shipped in

dry ice

shipped in

-

shipped in

ambient

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters