Skip to Content
Merck

Skip To

C4407

Human Collagen Type III

from human placenta, powder, suitable for cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32
EC Number:
232-697-4
MDL number:
Form:
powder
Assay:
≥90% (GE)
Biological source:
human placenta
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder

biological source

human placenta

Quality Segment

assay

≥90% (GE)

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

HIV, hepatitis B and hepatitis C, none detected

solubility

aqueous acid: soluble

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... COL3A1(1300)

Application

Collagen from human placenta has been used to assess the bioelectric effects of quinine on airway epithelial cells, to study the selective toxicity of engineered lentvirus lytic peptides and in a particle aggregation assay for the rapid detection of fibronectin, fibrinogen and collagen receptors on Staphylococcus aureus. It has also been used in a magnesium-dependent, collagen-binding assay during characterization of human lung tumor-associated antigens.

Preparation Note

This product was prepared by a modification of the pepsin extraction and salt fractionation method of Hill, R.J. and Harper, E., Anal. Biochem., 141 (1984). It is a lyophilized powder that can be reconstituted in sterile .5 M acetic acid, PBS or water at 1 mg/mL. A PBS solution will be stable for at least one year at -20°C.

Analysis Note

An SDS polyacrylamide gel electrophoresis test run under reducing conditions consistent with basement membrane collagen yields three major bands.

Other Notes

All collagen molecules are composed of three polypeptide chains arranged in a triple helical conformation, with a primary structure that is mostly a repeating motif with glycine in every third position and proline or 4-hydroxyproline frequently preceding the glycine residue.
Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Be wary of confusing Sigma-type designations with recognized collagen classification types.

Disclaimer

This product should be stored at 2-8 °C and will retain activity in these conditions for 2 years.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7774C3657C7521
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

biological source

human placenta

biological source

human placenta

biological source

human placenta

biological source

human placenta

assay

≥90% (GE)

assay

-

assay

-

assay

-

form

powder

form

powder

form

powder

form

powder

Gene Information

human ... COL3A1(1300)

Gene Information

bovine ... CSN2(281099), CSN3(281728)
human ... COL4A3(1285), COL4A4(1286), COL4A6(1288)

Gene Information

human ... COL5A1(1289)

Gene Information

human ... COL4A2(1284), COL4A3(1285), COL4A4(1286), COL4A5(1287), COL4A6(1288)

UniProt accession no.

Q03692

UniProt accession no.

P02666, P02668, P53420, Q01955, Q14031

UniProt accession no.

P20908

UniProt accession no.

P08572, P29400, P53420, Q01955, Q14031


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
C4407-1MG04061833492123
C4407-25MG04061833260999
C4407-5MG04061833492130

Questions

Reviews

No rating value

Active Filters