C-12375
Human Cardiac Fibroblasts (HCF)
500,000 cryopreserved cells
Synonym(s):
HCF cells
About This Item
Recommended Products
biological source
human heart (ventricles)
packaging
pkg of 500,000 cells
morphology
(fibroblast)
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cardiovascular diseases
shipped in
dry ice
storage temp.
−196°C
General description
Cell Line Origin
Application
Quality
Warning
Subculture Routine
Other Notes
Recommended products
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Yeast plasmid selection: Utilizes auxotrophic yeast strains for plasmid selection, enabling growth on synthetic media for better yield.
Yeast plasmid selection: Utilizes auxotrophic yeast strains for plasmid selection, enabling growth on synthetic media for better yield.
Overview of primary cells and media usage in biomedical culture applications highlights differences from cell lines.
Overview of primary cells and media usage in biomedical culture applications highlights differences from cell lines.
Related Content
Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.
Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.
Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.
Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service