Skip to Content
Merck
All Photos(2)

Key Documents

AV42146

Sigma-Aldrich

Anti-PPAP2A (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-LLP1a, Anti-LPP1, Anti-PAPα1, Anti-PAP-2a, Anti-PAP2, Anti-PAP2a2, Anti-PAP2alpha2, Anti-Phosphatidic acid phosphatase type 2A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

31 kDa

species reactivity

rat, mouse, bovine, human, dog, guinea pig, horse, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPAP2A(8611)

General description

Lipid phosphate phosphatases (LPP) dephosphorylate a variety of bioactive lipids including phosphatidate, lysophosphatidate, sphingosine 1-phosphate, and ceramide 1-phosphate.
The lipid phosphate phosphatase phosphatidic acid phosphatase type 2A/lipid phosphate phosphohydrolase type 1(PPAP2A, LLP1) dephosphorylates exogenous lysophosphatidate (LPA), a lipid mediator that stimulates cell proliferation and growth, and is involved in physiological and pathological processes such as wound healing, platelet activation, angiogenesis and the growth of tumours.

Specificity

Anti-PPAP2A (AB1) polyclonal antibody reacts with human, zebrafish, mouse, rat, chicken, bovine, pig, and rabbit phosphatidic acid phosphatase type 2A proteins.

Immunogen

Synthetic peptide directed towards the middle region of human PPAP2A

Application

Anti-PPAP2A (AB1) polyclonal antibody is used to tag phosphatidic acid phosphatase type 2A for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of phosphatidic acid phosphatase type 2A as an integral membrane bioactive lipid phosphatase.

Biochem/physiol Actions

PPAP2A is a member of the phosphatidic acid phosphatase (PAP) family. PAPs convert phosphatidic acid to diacylglycerol, and function in de novo synthesis of glycerolipids as well as in receptor-activated signal transduction mediated by phospholipase D. This protein is an integral membrane glycoprotein, and has been shown to be a surface enzyme that plays an active role in the hydrolysis and uptake of lipids from extracellular space. The expression of PPAP2A is found to be regulated by androgen in a prostatic adenocarcinoma cell line.The protein encoded by this gene is a member of the phosphatidic acid phosphatase (PAP) family. PAPs convert phosphatidic acid to diacylglycerol, and function in de novo synthesis of glycerolipids as well as in receptor-activated signal transduction mediated by phospholipase D. This protein is an integral membrane glycoprotein, and has been shown to be a surface enzyme that plays an active role in the hydrolysis and uptake of lipids from extracellular space. The expression of this gene is found to be regulated by androgen in a prostatic adenocarcinoma cell line. At least two alternatively spliced transcript variants encoding distinct isoforms have been described.

Sequence

Synthetic peptide located within the following region: DPDWSKINCSDGYIEYYICRGNAERVKEGRLSFYSGHSSFSMYCMLFVAL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Aranda et al.
Diabetologia, 56(6), 1444-1453 (2013-03-20)
The realisation that targeting agents in the vitreous is an effective approach to treating patients with diabetic retinopathy (DR) has increased awareness that changes in the composition/bioactivity of the vitreous is a contributor to the pathogenesis of DR. The overall
Ishita Chatterjee et al.
Cardiovascular research, 111(1), 105-118 (2016-04-30)
Lipid phosphate phosphatase-3 (LPP3) is expressed at high levels in endothelial cells (ECs). Although LPP3 is known to hydrolyse the phosphate group from lysolipids such as spingosine-1-phosphate and its structural homologues, the function of Lpp3 in ECs is not completely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service