Skip to Content
Merck
All Photos(1)

Key Documents

AV38012

Sigma-Aldrich

Anti-BACH1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-BTB and CNC homology 1, basic leucine zipper transcription factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

82 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BACH1(571)

General description

BTB-basic leucine zipper transcription factor is a member of a novel family of broad complex, tramtrack, bric-a-brac/poxvirus and zinc finger (BTB/POZ) basic region leucine zipper factors.
Bach1 functions as transcription repressor in transfection assays using fibroblast cells and as a transcriptional activator and repressor, respectively, in cultured erythroid cells. Bach1 is involved in the coordination of transcription activation and repression by Maf family members such as MafK. Bach1 (BTB and CNC homology 1, basic leucine zipper transcription factor 1) inhibits oxidative stress-inducible genes and is a negative regulator of oxidative stress-induced cellular senescence.

Specificity

Anti-BACH1 antibody reacts with human BTB and CNC homology 1, basic leucine zipper transcription factor 1 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human BACH1

Application

Anti-BACH1 antibody is used to tag BTB and CNC homology 1, basic leucine zipper transcription factor 1 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of basic leucine zipper transcription factor 1 in the regulation of processes such as cellular response to oxidative stress.

Biochem/physiol Actions

BACH1 is a transcription factor that belongs to the cap′n′collar type of basic region leucine zipper factor family (CNC-bZip). The encoded protein contains broad complex, tramtrack, bric-a-brac/poxvirus and zinc finger (BTB/POZ) domains, which is atypical of CNC-bZip family members. These BTB/POZ domains facilitate protein-protein interactions and formation of homo- and/or hetero-oligomers. When this encoded protein forms a heterodimer with MafK, it functions as a repressor of Maf recognition element (MARE) and transcription is repressed. Multiple alternatively spliced transcript variants have been identified for this gene. Some exons of this gene overlap with some exons from the C21orf41 gene, which is transcribed in an opposite orientation to this gene but does not seem to encode a protein.

Sequence

Synthetic peptide located within the following region: PPCARGNSEPGYARGQESQQMSTATSEQAGPAEQCRQSGGISDFCQQMTD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service