Skip to Content
Merck
All Photos(3)

Key Documents

A9451

Sigma-Aldrich

Aristolochic acid I sodium salt

powder, ≥97%

Synonym(s):

Aristolochic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H10NNaO7
CAS Number:
Molecular Weight:
363.25
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97%

form

powder

solubility

H2O: 50 mg/mL

SMILES string

[Na+].COc1cccc2c1cc([N+]([O-])=O)c3c(cc4OCOc4c23)C([O-])=O

InChI

1S/C17H11NO7.Na/c1-23-12-4-2-3-8-9(12)5-11(18(21)22)14-10(17(19)20)6-13-16(15(8)14)25-7-24-13;/h2-6H,7H2,1H3,(H,19,20);/q;+1/p-1

InChI key

BQVOPWJSBBMGBR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Component of some Chinese herbal medicines, and responsible for their nephrotoxicity. Prodrug activated by reduction of the nitro group to an amine, which forms cytotoxic DNA adducts.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marika Manolopoulou et al.
American journal of physiology. Renal physiology, 316(5), F847-F855 (2019-02-14)
Flow cytometry studies on injured kidney tubules are complicated by the low yield of nucleated single cells. Furthermore, cell-specific responses such as cell cycle dynamics in vivo have conventionally relied on indirect immunohistochemistry and proximal tubule markers that may be
Huan Lu et al.
Theranostics, 10(10), 4323-4333 (2020-04-16)
Rationale: Dietary exposure to aristolochic acids and similar compounds (collectively, AA) is a significant risk factor for nephropathy and subsequent upper tract urothelial carcinoma (UTUC). East Asian populations, who have a high prevalence of UTUC, have an unusual genome-wide AA-induced
Xiaoyan Wang et al.
Cell death & disease, 11(8), 620-620 (2020-08-17)
Acute kidney injury (AKI) has a critical role in the development of chronic kidney disease (CKD). Building on our previous findings, we explored the role of miR-382 in facilitating the transition of AKI to CKD using the Aristolochic acid (AA)
Khai Gene Leong et al.
Toxins, 13(10) (2021-10-23)
The plant-derived toxin, aristolochic acid (AA), is the cause of Chinese Herb Nephropathy and Balkan Nephropathy. Ingestion of high dose AA induces acute kidney injury, while chronic low dose ingestion leads to progressive kidney disease. Ingested AA is taken up
Sang Hoon Kim et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(18), 8440-8445 (2010-04-21)
Mammalian sweet, bitter, and umami taste is mediated by a single transduction pathway that includes a phospholipase C (PLC)beta and one cation channel, TRPM5. However, in insects such as the fruit fly, Drosophila melanogaster, it is unclear whether different tastants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service