Skip to Content
Merck
All Photos(1)

Key Documents

A8438

Sigma-Aldrich

Anti-Rat IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Goat Anti-Rat IgG (whole molecule)–AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

technique(s)

direct ELISA: 1:30,000
immunohistochemistry (frozen sections): 1:50
western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG, a monomer, is the predominant antibody circulating in blood which assist in phagocytic destruction of foreign microorganism. Anti-rat IgG (whole molecule)-alkaline phosphatase antibody (diluted 1: 30 000) can be used in rat MPO (myeloperoxidase) ELISA. It may also use as secondary antibody in immunodetection method. Anti-rat IgG (whole molecule)-alkaline phosphatase antibody reacts specifically with all rat immunoglobulins.

Immunogen

Purified rat IgG.

Application

Anti-rat IgG (whole molecule)–alkaline phosphatase antibody can be used in ELISA and western blot assays.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Western blot was performed to analyze purified proteins over expressed in E.coli using alkaline-phosphatase-conjugated goat anti-rat IgG at a 1:1000 dilution. Western blot was developed using 5-bromo-4-chloro-3indolyl phosphate/nitro blue tetrazolium tablets (Sigma).

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2, 10 mM glycine, 50% glycerol and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianmin Zuo et al.
Journal of virology, 82(5), 2385-2393 (2007-12-21)
The DNase/alkaline exonuclease (AE) genes are well conserved in all herpesvirus families, but recent studies have shown that the AE proteins of gammaherpesviruses such as Epstein-Barr virus (EBV) and Kaposi's sarcoma-associated herpesvirus (KSHV) exhibit an additional function which shuts down
Kailash C Pandey et al.
The Journal of biological chemistry, 279(5), 3484-3491 (2003-11-20)
The Plasmodium falciparum cysteine protease falcipain-2 is a trophozoite hemoglobinase and potential antimalarial drug target. Unlike other studied papain family proteases, falcipain-2 does not require its prodomain for folding to active enzyme. Rather, folding is mediated by an amino-terminal extension
Chandra Ramakrishnan et al.
Scientific reports, 9(1), 1474-1474 (2019-02-08)
Almost any warm-blooded creature can be an intermediate host for Toxoplasma gondii. However, sexual reproduction of T. gondii occurs only in felids, wherein fertilisation of haploid macrogametes by haploid microgametes, results in diploid zygotes, around which a protective wall develops
Joya E Nahon et al.
Atherosclerosis, 281, 47-55 (2019-01-19)
Stabilin-1 (STAB1) is a scavenger receptor expressed on alternatively activated macrophages and sinusoidal endothelial cells. Its ligands include oxidized low-density lipoprotein (LDL) and the extracellular matrix glycoprotein SPARC and it is present in both human and murine atherosclerotic lesions. We
Frank Lennartz et al.
Nature communications, 9(1), 3822-3822 (2018-09-22)
The quest to develop an effective malaria vaccine remains a major priority in the fight against global infectious disease. An approach with great potential is a transmission-blocking vaccine which induces antibodies that prevent establishment of a productive infection in mosquitos

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service