Skip to Content
Merck
All Photos(1)

Documents

A2201

Sigma-Aldrich

Amyloid β-Protein Fragment 35-25

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H81N13O14S
CAS Number:
Molecular Weight:
1060.27
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)CNC(=O)C(CC(C)C)NC(=O)C(N)CCSC)C(=O)NC(C(C)CC)C(=O)NC(C)C(=O)NCC(=O)NC(CCCCN)C(=O)NC(CC(N)=O)C(=O)NC(CO)C(=O)NCC(O)=O

InChI

1S/C45H81N13O14S/c1-9-24(5)36(57-34(62)20-50-40(67)29(17-23(3)4)54-39(66)27(47)14-16-73-8)45(72)58-37(25(6)10-2)44(71)52-26(7)38(65)49-19-33(61)53-28(13-11-12-15-46)42(69)55-30(18-32(48)60)43(70)56-31(22-59)41(68)51-21-35(63)64/h23-31,36-37,59H,9-22,46-47H2,1-8H3,(H2,48,60)(H,49,65)(H,50,67)(H,51,68)(H,52,71)(H,53,61)(H,54,66)(H,55,69)(H,56,70)(H,57,62)(H,58,72)(H,63,64)

InChI key

IDGOADDOQWKZOX-UHFFFAOYSA-N

Gene Information

human ... APP(351)

Amino Acid Sequence

Met-Leu-Gly-Ile-Ile-Ala-Gly-Lys-Asn-Ser-Gly

General description

Amyloid β precursor protein (APP) is a type-1 transmembrane protein. APP is expressed widely and mainly found in the synapses and neurons. The APP gene is mapped to human chromosome 21q21.3.

Application

Amyloid β-Protein Fragment 35-25 has been used as a control peptide for Aβ-induced apoptosis to study the role of hepcidin overexpression in astrocytes against amyloid-β induced brain damage in mice.

Biochem/physiol Actions

Proteolytic processing of amyloid β precursor protein (APP) forms the β amyloid protein (Aβ) as part of the brain amyloid plaques which are the key biomarkers in Alzheimer′s disease. APP is known to be associated with neurogenesis, synapse formation, axonal transport, neuronal migration and neurite outgrowth. It is also related to mitochondria-associated endoplasmic reticulum membranes (MAMs) activity, antimicrobial protection, cholesterol, and iron homeostasis. Amyloid β-Protein Fragment 35-25 is an inactive control.

Other Notes

The N—C inverted sequence of fragment 25-35

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Anatoliy I Yashin et al.
Mechanisms of ageing and development, 196, 111477-111477 (2021-04-03)
Emerging evidence from experimental and clinical research suggests that stress-related genes may play key roles in AD development. The fact that genome-wide association studies were not able to detect a contribution of such genes to AD indicates the possibility that
Jasmine Chebli et al.
Scientific reports, 11(1), 19115-19115 (2021-09-29)
Amyloid precursor protein (APP) is expressed in many tissues in human, mice and in zebrafish. In zebrafish, there are two orthologues, Appa and Appb. Interestingly, some cellular processes associated with APP overlap with cilia-mediated functions. Whereas the localization of APP
Elvis Cuevas et al.
Metabolic brain disease, 34(5), 1365-1374 (2019-07-04)
The amyloid β-peptide (Aβ) is transported across the blood-brain barrier (BBB) by binding with the receptor for advanced glycation end products (RAGE). Previously, we demonstrated that the Aβ fraction 25-35 (Aβ25-35) increases RAGE expression in the rat hippocampus, likely contributing
Alzheimer's disease: amyloid beta-peptide antibody vaccine as plaque remover.
A Kumar
Journal of biosciences, 25(4), 315-316 (2000-12-20)
Xinwei Zhang et al.
Cell death discovery, 6(1), 113-113 (2020-12-11)
Progressive iron accumulation in the brain and iron-induced oxidative stress are considered to be one of the initial causes of Alzheimer's disease (AD), and modulation of brain iron level shows promise for its treatment. Hepcidin expressed by astrocytes has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service