Skip to Content
Merck

Skip To

92897

Poly(ethylene glycol)

greener alternative

BioUltra, 10,000

Synonym(s):

Polyethylene glycol, Polyethylene oxide, Polyoxyethylene, PEG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352104
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

BioUltra

form

solid

mol wt

Mr 8500-11500

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

insoluble matter:, passes filter test, ≤0.001% peroxides (as H2O2):

pH

5.5-7.0 (25 °C, 50 mg/mL in H2O)

mp

62-65 °C

solubility

H2O: 50 mg/mL at 25 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

50 mg/mL in H2O

UV absorption

λ: 260 nm Amax: 0.09, λ: 280 nm Amax: 0.03

greener alternative category

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

General description

Poly(ethylene glycol) (PEG), a linear polyether of ethylene glycol (ethane-1,2-diol), is soluble in water, benzene, ethanol, acetonitrile, and dichloromethane. However, it is insoluble in diethyl ether and hexane. This non-ionic polymer[1] can exist in the monomer and aqueous phases.[2]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. Polyethylene glycol (PEG) is an eco-friendly, biodegradable polymer widely used in pharmaceuticals and cosmetics. Its non-toxic nature and versatility make it a sustainable choice, derived from renewable resources, contributing to greener product formulations. Click here for more information.

Application

Poly(ethylene glycol) has been used:

  • in cryopreservation of cell-laden multi-assay platforms (MAPs)[3]
  • as a macromolecular crowder to study its effects on fluorescein isothiocyanate (FITC)-conjugated type-I collagen[4]
  • as a model surfactant in surfactant formulations to evaluate the interfacial aggregation of antibody formulations in response to interfacial stress[5]

Biochem/physiol Actions

Poly(ethylene glycol) (PEG) is used in several applications including medicine. This polymer occurs as a preservative and is used in biomedical applications like drug delivery systems, biosensing, and tissue engineering. PEG is also used in cosmetic ointments and creams. It is used as a binding and dispersing agent in chemical and other industries.

Features and Benefits

  • Environment friendly[2]
  • Biocompatible[2]
  • Unique solvent solution[2]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
882768951081255
form

solid

form

powder

form

solid

form

solid

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

mol wt

Mr 8500-11500

mol wt

Mr 3000-3700

mol wt

Mr 7000-9000

mol wt

Mr 5000-7000

cation traces

Al: ≤5 mg/kg, Ba: ≤5 mg/kg, Ca: ≤10 mg/kg, Co: ≤5 mg/kg, Cu: ≤5 mg/kg, K: ≤200 mg/kg, Mg: ≤5 mg/kg, Mo: ≤5 mg/kg, Ni: ≤5 mg/kg, Sr: ≤5 mg/kg, As: ≤0.1 mg/kg, Cd: ≤5 mg/kg, Fe: ≤5 mg/kg, Mn: ≤5 mg/kg, Pb: ≤5 mg/kg, Bi: ≤5 mg/kg, Li: ≤5 mg/kg, Zn: ≤5 mg/kg, Cr: ≤5 mg/kg, Na: ≤200 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤5 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

mp

62-65 °C

mp

-

mp

61-64 °C

mp

-

impurities

insoluble matter:, passes filter test, ≤0.001% peroxides (as H2O2):

impurities

insoluble matters, passes filter test

impurities

insoluble matter:, passes filter test, ≤0.001% peroxides (as H2O2):

impurities

insoluble matter, passes filter test, ≤20 mg/kg peroxides (as H2O2)


Still not finding the right product?

Explore all of our products under Poly(ethylene glycol)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Polymers for 3D bioprinting
Beata K
Journal of Separation Science (2022)
Polymers and polymeric hybrids for targeted drug delivery
Islam R and Fang J
Journal of Separation Science, 303-322 (2022)
Aadithya Kannan et al.
Journal of colloid and interface science, 550, 128-138 (2019-05-06)
Monoclonal antibodies (mAbs) are therapeutic proteins used in the treatment of many diseases due to their specificity in binding targets. Aggregation of these molecules is a major challenge in their formulation development. MAbs spontaneously adsorb onto air-solution interfaces and experience



Global Trade Item Number

SKUGTIN
92897-5G-KC-F04061826546277
92897-1KG-F04061826715420
92897-250G-F04061833270516

Questions

Reviews

No rating value

Active Filters