Skip to Content
Merck
All Photos(1)

Documents

30120

Sigma-Aldrich

L-Cysteine hydrochloride

≥99.0% (RT)

Synonym(s):

L-Cysteine hydrochloride (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)COOH · HCl
CAS Number:
Molecular Weight:
157.62
Beilstein:
3560277
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

product name

L-Cysteine hydrochloride, anhydrous, ≥99.0% (RT)

Assay

≥99.0% (RT)

form

powder or crystals

optical activity

[α]20/D +6.4±0.3°, c = 5% in 5 M HCl

impurities

≤0.5% foreign amino acids

ign. residue

≤0.1% (as SO4)

loss

≤1.0% loss on drying

color

white

mp

174.1-174.7 °C

solubility

H2O: 1 g/10 mL, clear, colorless

anion traces

sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤500 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

peptide synthesis

SMILES string

Cl[H].N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S.ClH/c4-2(1-7)3(5)6;/h2,7H,1,4H2,(H,5,6);1H/t2-;/m0./s1

InChI key

IFQSXNOEEPCSLW-DKWTVANSSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Drug delivery systems: A study utilized L-Cysteine HCl in the formulation of mixed micelles for the enhanced delivery of carvedilol, a cardiovascular drug, illustrating its utility in improving solubility and bioavailability of pharmaceutical compounds (Öztürk et al., 2022).

Biochem/physiol Actions

NMDA glutamatergic receptor agonist that is also an agonist at AMPA glutamatergic receptors at high concentrations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Omicron BQ.1.1 and XBB.1 unprecedentedly escape broadly neutralizing antibodies elicited by prototype vaccination.
Ju, et al.
Cell Reports, 42, 112532-112532 (2023)
Orit Redy-Keisar et al.
Nature protocols, 9(1), 27-36 (2013-12-07)
This protocol describes the synthesis of modular turn-ON QCy7-based probes for the detection of biologically relevant analytes, such as hydrogen peroxide, ubiquitous sulfhydryl and β-galactosidase. The probes presented herein are prepared through a simple procedure that involves the preliminary alkylation
Kathleen R Stewart-Morgan et al.
Nature cell biology, 25(1), 183-193 (2023-01-13)
DNA methylation is a critical epigenetic mark in mammalian cells. Many aspects of DNA methylation maintenance have been characterized; however, the exact kinetics of post-replicative methylation maintenance remain a subject of debate. Here we develop isolation of DNA by 5-ethynyl-deoxyuridine
Michela Miani et al.
Cell metabolism, 28(4), 557-572 (2018-07-19)
The gut microbiota is essential for the normal function of the gut immune system, and microbiota alterations are associated with autoimmune disorders. However, how the gut microbiota prevents autoimmunity in distant organs remains poorly defined. Here we reveal that gut
Laura Glorieux et al.
International journal of molecular sciences, 24(12) (2023-06-28)
The pancreas is a complex organ consisting of differentiated cells and extracellular matrix (ECM) organized adequately to enable its endocrine and exocrine functions. Although much is known about the intrinsic factors that control pancreas development, very few studies have focused

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service