Skip to Content
Merck

14366C

EX-CELL® Advanced CHO Fed-batch Medium

liquid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.75
UNSPSC Code:
41161501
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

sterile-filtered

Quality Segment

product line

EX-CELL®

form

liquid

feature

subscription eligible

technique(s)

cell culture | mammalian: suitable (suspension)

components

L-glutamine: no

shipped in

ambient

storage temp.

2-8°C

General description

EX-CELL® AdvancedCHO Fed-batch Medium is chemically-defined and contains no animal derived components. This platform contains no hypoxanthine andthymidine, L-glutamine, phenol red or 2-mercaptoethanol (2-ME). This medium isintended to be a part of a fed-batch platform and used with EX-CELL® AdvancedCHO Feed 1 (24367C - with glucose or 24368C - without glucose).

Application

EX-CELL® Advanced CHO Fed-batch Medium from SAFC has been specifically developed to support growth and productivity across a diverse set of Chinese Hamster Ovary (CHO) cells in fed-batch cultures while still allowing for flexibility in the adjustment of protein quality attributes.

This contemporary fed-batch platform was developed using a strategy of Multivariate Analysis to establish correlations in which the formulation components influence critical process and product attributes such as titer, fold expansion, receptor expression, and glycosylation.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Donot freeze. Protect from light. Several components of the medium arelight labile and should not be exposed to light for lengthy periods of time.


Still not finding the right product?


wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library