Skip to Content
Merck
All Photos(1)

Key Documents

11635

Sigma-Aldrich

Azomethine-H monosodium salt hydrate

p.a.

Synonym(s):

4-Hydroxy-5-(2-hydroxybenzylideneamino)-naphthalene-2,7-disulfonic acid monosodium salt hydrate, 4-Hydroxy-5-(salicylideneamino)-2,7-naphthalenedisulfonic acid monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12NNaO8S2 · xH2O
CAS Number:
Molecular Weight:
445.40 (anhydrous basis)
Beilstein:
4100175
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

p.a.

form

solid

technique(s)

UV/Vis spectroscopy: suitable

SMILES string

O.[Na+].Oc1ccccc1\C=N\c2cc(cc3cc(cc(O)c23)S(O)(=O)=O)S([O-])(=O)=O

InChI

1S/C17H13NO8S2.Na.H2O/c19-15-4-2-1-3-10(15)9-18-14-7-12(27(21,22)23)5-11-6-13(28(24,25)26)8-16(20)17(11)14;;/h1-9,19-20H,(H,21,22,23)(H,24,25,26);;1H2/q;+1;/p-1/b18-9+;;

InChI key

AAIGDXDVSZJVSW-IFJQNBRBSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Azomethine-H monosodium salt hydrate is a complexing agent used to determine the concentration of boron in the aqueous solutions by the colorimetric method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service