Skip to Content
Merck
All Photos(2)

Key Documents

O4504

Sigma-Aldrich

2-Octanol

97%

Synonym(s):

sec-Caprylic alcohol, Capryl alcohol, Ethyl-N-amylcarbinol, Ethylpentylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH3
CAS Number:
Molecular Weight:
130.23
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21
Assay:
97%
bp:
174-181 °C (lit.)
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

liquid

expl. lim.

0.34-6.3 %

refractive index

n20/D 1.426 (lit.)

bp

174-181 °C (lit.)

mp

-38 °C

density

0.819 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC(C)O

InChI

1S/C8H18O/c1-3-4-5-6-7-8(2)9/h8-9H,3-7H2,1-2H3

InChI key

SJWFXCIHNDVPSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Octanol is a secondary alcohol that can be oxidized by nitric acid to form 2-octanone.[1][2] The performance of Al-Cu and Al-Cu-Mg mixed oxides to catalyze the dehydrogenation of 2-octanol has been investigated.[3] The reaction kinetics of the formation of ammonium sec-octyl sulfate by reaction of 2-octanol with sulfamic acid has been reported.[4]

Application

2-Octanol may be used as a reducer in the preparation of monodispersed nickel nanocrystals.[5]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Size-controlled monodispersed nickel nanocrystals using 2-octanol as reducing agent.
Huaman JLC, et al.
CrystEngComm, 15(4), 729-737 (2013)
Runaway behavior and thermally safe operation of multiple liquid-liquid reactions in the semi-batch reactor: The nitric acid oxidation of 2-octanol.
Van Woezik BAA and Westerterp KR.
Chemical Engineering and Processing, 41(1), 59-77 (2002)
A Study of the Kinetics of the Sulfation of 2-Octanol by Sulfamic Acid.
Ho WH.
J. Chin. Chem. Soc., 34(4), 257-261 (1987)
The nitric acid oxidation of 2-octanol. A model reaction for multiple heterogeneous liquid-liquid reactions.
Van Woezik BAA and Westerterp KR.
Chemical Engineering and Processing, 39(6), 521-537 (2000)
Characterization of Al-Cu and Al-Cu-Mg mixed oxides and their catalytic activity in dehydrogenation of 2-octanol.
Crivello M, et al.
Catalysis Today, 107, 215-222 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service