Skip to Content
Merck
All Photos(4)

Key Documents

M7634

Sigma-Aldrich

Manganese(II) sulfate monohydrate

ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnSO4 · H2O
CAS Number:
Molecular Weight:
169.02
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥99%
form:
powder

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

powder

pH

3-3.5 (20 °C, 50 g/L)

storage temp.

room temp

SMILES string

O.[Mn++].[O-]S([O-])(=O)=O

InChI

1S/Mn.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

ISPYRSDWRDQNSW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Manganese(II) sulfate monohydrate (MnSO4·H2O) is a hydrated manganese salt. The neurological response caused in rhesus monkeys due to MnSO4 exposure have been studied. The limiting molar conductance and association constant of MnSO4 dissolved in aqueous N,N-dimethylformamide have been determined. The solubility, binary interaction parameters and solubility parameters were obtained for aqueous solution containing manganese sulfate monohydrate and magnesium sulfate.

Application

Manganese(II) sulfate monohydrate may be used to assist the oxidation of arenes and sulfides by potassium permanganate.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

David C Dorman et al.
Toxicological sciences : an official journal of the Society of Toxicology, 92(1), 201-210 (2006-04-21)
High-dose human exposure to manganese results in manganese accumulation in the basal ganglia and dopaminergic neuropathology. Occupational manganese neurotoxicity is most frequently linked with manganese oxide inhalation; however, exposure to other forms of manganese may lead to higher body burdens.
Green oxidations. Manganese(II) sulfate aided oxidations of organic compounds by potassium permanganate.
Shaabani A, et al.
Monatshefte fur Chemie / Chemical Monthly, 138(7), 649-651 (2007)
Conductance studies on manganese(II), cobalt(II), nickel(II), and cadmium(II) sulfates in water+ N,N-dimethylformamide mixtures at 293.15 K.
Molinou IE and Tsierkezos NG.
Journal of Chemical and Engineering Data, 46(6), 1399-1403 (2001)
Solubility of Manganese Sulfate Monohydrate in the Presence of Trace Quantities of Magnesium Sulfate Heptahydrate in Water.
Maeda K, et al.
Developments in Chemical Engineering and Mineral Processing, 11(5-6), 423-435 (2003)
Saleem Alameh et al.
PLoS pathogens, 16(8), e1008836-e1008836 (2020-09-01)
Anthrax is a major zoonotic disease of wildlife, and in places like West Africa, it can be caused by Bacillus anthracis in arid nonsylvatic savannahs, and by B. cereus biovar anthracis (Bcbva) in sylvatic rainforests. Bcbva-caused anthrax has been implicated

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service