Skip to Content
Merck
All Photos(1)

Key Documents

G7893

Sigma-Aldrich

Glycerol

Glycerol

ACS reagent, ≥99.5%

Synonym(s):

1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.05
grade:
ACS reagent
Assay:
≥99.5%
technique(s):
electrophoresis: suitable
bp:
182 °C/20 mmHg (lit.)
vapor pressure:
<1 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99.5%

form

viscous liquid

autoignition temp.

698 °F

expl. lim.

2.6-11.3 %

technique(s)

electrophoresis: suitable

impurities

Acrolein and glucose, passes test
Fatty acid esters, passes test
H2SO4, passes test (darkened)
Neutrality, passes test
≤0.003% Chlorinated compounds (as Cl)
≤0.500% Water (H2O)

ign. residue

≤0.0050%

color

APHA: ≤10

refractive index

n20/D 1.474 (lit.)

pH

5.5-8

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

anion traces

sulfate (SO42-): passes test

cation traces

heavy metals (as Pb): ≤2 ppm

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycerol, also known as Glycerin, is a sugar alcohol. It is a viscous liquid that is odorless, colorless, and hygroscopic in nature. It is highly soluble in water and can also react with aldehydes, ketones, acetic acid, methyl, and ethyl acetates forming acetals, cetals, and glycerine acetates. It finds numerous applications in the food, pharmaceutical, and cosmetic sectors.[1]

Application

Glycerol is used as a starting material or reactant in chemical synthesis.

greener alternative product

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycerol from biodiesel production: the new corn for dairy cattle
S Shawn and Donkin
Revista Brasileira de Zootecnia, 37, 280-286 (2008)
Hovsep Mahdessian et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(24), 8913-8918 (2014-06-14)
Genome-wide association studies have identified a locus on chromosome 19 associated with plasma triglyceride (TG) concentration and nonalcoholic fatty liver disease. However, the identity and functional role of the gene(s) responsible for these associations remain unknown. Of 19 expressed genes
Shelley L Forrest et al.
The Journal of comparative neurology, 522(17), 3900-3927 (2014-07-22)
Artemin is a member of the glial cell line-derived neurotrophic factor (GDNF) family that has been strongly implicated in development and regeneration of autonomic nerves and modulation of nociception. Whereas other members of this family (GDNF and neurturin) primarily target
Benoît Ranchoux et al.
The American journal of pathology, 185(2), 356-371 (2014-12-17)
Pulmonary veno-occlusive disease (PVOD) is an uncommon form of pulmonary hypertension (PH) characterized by progressive obstruction of small pulmonary veins and a dismal prognosis. Limited case series have reported a possible association between different chemotherapeutic agents and PVOD. We evaluated
Marc Simard et al.
The Journal of clinical endocrinology and metabolism, 100(1), E129-E139 (2014-10-17)
Corticosteroid-binding globulin (CBG) is encoded by SERPINA6. It is the major plasma binding protein of glucocorticoids and regulates plasma cortisol levels and bioavailability in humans. Several proteases target CBG and disrupt its steroid-binding properties. To date, most genetic deficiencies that

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the viscosity of glycerol?

    1 answer
    1. The viscosity of glycerol is 648 Centistokes at 20 Deg C (68 Deg F).

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service