Skip to Content
Merck
All Photos(1)

Key Documents

61197

Sigma-Aldrich

Copper(II) nitrate trihydrate

purum p.a., 98.0-103% (RT)

Synonym(s):

Cupric nitrate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(NO3)2 · 3H2O
CAS Number:
Molecular Weight:
241.60
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

Assay

98.0-103% (RT)

form

powder, crystals or chunks

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Cu.2NO3.3H2O/c;2*2-1(3)4;;;/h;;;3*1H2/q+2;2*-1;;;

InChI key

SXTLQDJHRPXDSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Copper(II) nitrate trihydrate is a hydrated copper salt. It participates as catalyst in the solvent-free synthesis of 3-aminopropenones and 3-aminopropenoates.

Application

Copper(II) nitrate trihydrate may be used in the preparation of the following mononuclear copper(II) complexes (phen = 1,10-phenanthroline, dpq = dipyrido[3,2-d:2′,3′-f]quinoxaline and dppz = dipyrido[3,2-a:2′,3′-c]phenazine):
  • [Cu(dpq)(NO3)2]
  • [Cu(dpq)(NO3)(H2O)2](NO3)
  • [Cu(dpq)(NO3)2(H2O)2]·2H2O
  • [Cu(dppz)(NO3)2(H2O)]·H2O

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple and efficient synthesis of 3-aminopropenones and 3-aminopropenoates catalyzed by copper (II) nitrate trihydrate under solvent-free conditions.
Li GC.
Monatshefte fur Chemie / Chemical Monthly, 139(7), 789-792 (2008)
Anitha M Thomas et al.
Journal of inorganic biochemistry, 94(1-2), 171-178 (2003-03-07)
A series of mononuclear copper(II) complexes having a 1:1 molar ratio of copper and the planar heterocyclic base like 1,10-phenanthroline (phen), dipyrido[3,2-d:2',3'-f]quinoxaline (dpq) and dipyrido[3,2-a:2',3'-c]phenazine (dppz) are prepared from a reaction of copper(II) nitrate.trihydrate and the base (L) in ethanol
Adetayo M Mustapha et al.
Journal of mass spectrometry : JMS, 50(1), 145-152 (2015-01-21)
Efforts to improve agricultural productivity have led to a growing dependency on organophosphorus pesticides. Phosphorothioate and phosphorodithioate pesticides are organophosphorus pesticide subclasses with widespread application for the control of insects feeding on vegetables and fruits. However, even low doses of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service