Skip to Content
Merck
All Photos(1)

Key Documents

27225

Sigma-Aldrich

Acetic acid

glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%

Synonym(s):

Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

Agency

meets purity specifications of USP (identity)

Quality Level

vapor density

2.07 (vs air)

grade

puriss.

Assay

99.8-100.5%

form

liquid

autoignition temp.

800 °F

quality

meets analytical specification of Ph. Eur., BP, USP, FCC

expl. lim.

16 %, 92 °F
4 %, 59 °F

technique(s)

mass spectrometry (MS): suitable

impurities

K2Cr2O7 red. substances, complies
residual solvents, complies
≤0.0002% heavy metals (as Pb)
≤0.005% non-volatile matter
≤0.01% KMnO4 red. matter (as HCOOH)

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50 g/L)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

transition temp

freezing point ≥15.6 °C

density

1.04 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤2 mg/kg
sulfate (SO42-): ≤5 mg/kg

cation traces

Al: ≤0.5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤2 mg/kg
Pb: ≤0.5 mg/kg
Zn: ≤5 mg/kg

suitability

corresponds for appearance

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetic acid is an organic carboxylic acid. It is a hygroscopic liquid having pungent odor. Its anhydrous form is commonly referred as glacial acetic acid. It forms dimers linked by hydrogen bonds in solution and gaseous phases. It is weakly acidic in nature.

Application

Acetic acid is widely employed as solvent for various oxidation reactions. It may be used in the analysis of synthetic peptides by electron transfer dissociation (ETD) tandem mass spectrometry (MS/MS). It may be used in the preparation of peroxyacetic acid (disinfectant), via reaction with perhydrol (30% hydrogen peroxide).

Other Notes

The article number 27225-4X2.5L-M will be discontinued. Please order the single bottle 27225-2.5L-M which is physically identical with the same exact specifications.
The article number 27225-4X2.5L-R will be discontinued. Please order the single bottle 27225-2.5L-R which is physically identical with the same exact specifications.
The article number 27225-6X1L-M will be discontinued. Please order the single bottle 27225-1L-M which is physically identical with the same exact specifications.
The article number 27225-6X1L-R will be discontinued. Please order the single bottle 27225-1L-R which is physically identical with the same exact specifications.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John E P Syka et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(26), 9528-9533 (2004-06-24)
Peptide sequence analysis using a combination of gas-phase ion/ion chemistry and tandem mass spectrometry (MS/MS) is demonstrated. Singly charged anthracene anions transfer an electron to multiply protonated peptides in a radio frequency quadrupole linear ion trap (QLT) and induce fragmentation
Eagleson M.
Concise Encyclopedia Chemistry, 5-5 (1994)
Isabelle Brunet et al.
The Journal of clinical investigation, 124(7), 3230-3240 (2014-06-18)
Autonomic sympathetic nerves innervate peripheral resistance arteries, thereby regulating vascular tone and controlling blood supply to organs. Despite the fundamental importance of blood flow control, how sympathetic arterial innervation develops remains largely unknown. Here, we identified the axon guidance cue
Katharine Trenholme et al.
Antimicrobial agents and chemotherapy, 58(7), 3666-3678 (2014-04-16)
Therapies to prevent transmission of malaria parasites to the mosquito vector are a vital part of the global malaria elimination agenda. Primaquine is currently the only drug with such activity; however, its use is limited by side effects. The development
Grete Andersen et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 308(2), R123-R130 (2014-11-21)
In healthy individuals, postexercise protein supplementation increases muscle protein anabolism. In patients with muscular dystrophies, aerobic exercise improves muscle function, but the effect of exercise on muscle protein balance is unknown. Therefore, we investigated 1) muscle protein balance before, during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service